Assay Details
| Gene Symbol: LILRA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021250.3 | NP_067073.1 | 2 - 3 | 208 | 86 | |
| NM_181879.2 | NP_870994.1 | 2 - 3 | 208 | 86 | ||
| GenBank mRNA | AF324830.1 | - | 2 - 3 | 208 | 86 | |
| AF499916.1 | - | 2 - 3 | 151 | 86 | ||
| AF499918.1 | - | 2 - 3 | 167 | 86 | ||
| EU915611.1 | - | 2 - 3 | 148 | 86 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19002719.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19025039.hg.1 | TC19002719.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19025039.hg.1 | ENST00000301219.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19025039.hg.1 | ENST00000346508.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19025039.hg.1 | ENST00000432233.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19025039.hg.1 | ENST00000446712.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19025039.hg.1 | NM_021250.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19025039.hg.1 | NM_181879.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19025039.hg.1 | NM_181985.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19025039.hg.1 | NM_181986.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19025040.hg.1 | TC19002719.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19025040.hg.1 | ENST00000301219.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19025040.hg.1 | ENST00000432233.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19025040.hg.1 | NM_021250.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19025040.hg.1 | NM_181879.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | ENST00000432233.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | ENST00000432233.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | ENST00000432233.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | ENST00000432233.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | ENST00000432233.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | ENST00000432233.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | ENST00000432233.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | ENST00000432233.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | ENST00000432233.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | ENST00000432233.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | ENST00000432233.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | ENST00000432233.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | ENST00000446712.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | ENST00000446712.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | ENST00000446712.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | ENST00000446712.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | ENST00000446712.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | ENST00000446712.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | ENST00000446712.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | ENST00000446712.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | ENST00000477720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | ENST00000477720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | ENST00000477720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | ENST00000477720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | ENST00000486742.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | ENST00000486742.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | ENST00000486742.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | ENST00000486742.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | ENST00000486742.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | ENST00000486742.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | ENST00000486742.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | ENST00000486742.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | ENST00000486742.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | ENST00000489504.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | ENST00000489504.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | ENST00000489504.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | ENST00000489504.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | ENST00000489504.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | ENST00000489504.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | ENST00000489504.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | ENST00000489504.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | NM_021250.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | NM_021250.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | NM_021250.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | NM_021250.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | NM_021250.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | NM_021250.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | NM_021250.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | NM_021250.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | NM_021250.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | NM_021250.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | NM_021250.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | NM_021250.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | NM_181879.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | NM_181879.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | NM_181879.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | NM_181879.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | NM_181879.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | NM_181879.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | NM_181879.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | NM_181879.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | NM_181879.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | NM_181879.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | NM_181985.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | NM_181985.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | NM_181985.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | NM_181985.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | NM_181985.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | NM_181985.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | NM_181985.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | NM_181985.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | NM_181985.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | NM_181985.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | NM_181985.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | NM_181986.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | NM_181986.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | NM_181986.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | NM_181986.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | NM_181986.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | NM_181986.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | NM_181986.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | NM_181986.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900012040.hg.1 | NM_181986.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900208064.hg.1 | ENST00000432233.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900208064.hg.1 | ENST00000446712.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900208064.hg.1 | NM_021250.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900208064.hg.1 | NM_181879.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900208064.hg.1 | TC1900012040.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900208064.hg.1 | ENST00000432233.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900208064.hg.1 | ENST00000446712.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900208064.hg.1 | NM_021250.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900208064.hg.1 | NM_181879.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | ENST00000432233.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | ENST00000432233.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | ENST00000432233.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | ENST00000432233.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | ENST00000432233.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | ENST00000432233.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | ENST00000432233.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | ENST00000432233.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | ENST00000432233.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | ENST00000432233.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | ENST00000432233.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | ENST00000432233.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | ENST00000446712.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | ENST00000446712.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | ENST00000446712.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | ENST00000446712.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | ENST00000446712.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | ENST00000446712.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | ENST00000446712.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | ENST00000446712.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | ENST00000477720.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | ENST00000477720.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | ENST00000477720.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | ENST00000477720.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | ENST00000486742.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | ENST00000486742.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | ENST00000486742.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | ENST00000486742.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | ENST00000486742.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | ENST00000486742.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | ENST00000486742.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | ENST00000486742.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | ENST00000486742.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | ENST00000489504.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | ENST00000489504.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | ENST00000489504.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | ENST00000489504.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | ENST00000489504.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | ENST00000489504.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | ENST00000489504.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | ENST00000489504.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | NM_021250.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | NM_021250.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | NM_021250.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | NM_021250.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | NM_021250.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | NM_021250.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | NM_021250.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | NM_021250.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | NM_021250.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | NM_021250.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | NM_021250.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | NM_021250.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | NM_181879.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | NM_181879.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | NM_181879.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | NM_181879.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | NM_181879.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | NM_181879.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | NM_181879.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | NM_181879.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | NM_181879.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | NM_181879.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | NM_181985.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | NM_181985.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | NM_181985.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | NM_181985.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | NM_181985.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | NM_181985.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | NM_181985.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | NM_181985.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | NM_181985.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | NM_181985.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | NM_181985.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | NM_181986.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | NM_181986.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | NM_181986.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | NM_181986.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | NM_181986.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | NM_181986.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | NM_181986.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | NM_181986.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900012040.hg.1 | NM_181986.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900208063.hg.1 | ENST00000432233.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900208063.hg.1 | ENST00000446712.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900208063.hg.1 | ENST00000486742.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900208063.hg.1 | ENST00000489504.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900208063.hg.1 | NM_021250.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900208063.hg.1 | NM_181879.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900208063.hg.1 | NM_181985.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900208063.hg.1 | NM_181986.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900208063.hg.1 | TC1900012040.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900208063.hg.1 | ENST00000432233.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900208063.hg.1 | ENST00000446712.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900208063.hg.1 | ENST00000486742.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900208063.hg.1 | ENST00000489504.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900208063.hg.1 | NM_021250.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900208063.hg.1 | NM_181879.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900208063.hg.1 | NM_181985.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900208063.hg.1 | NM_181986.2 | Refseq |

LILRA5
Genomic Map