Assay Details
Gene Symbol: C19orf57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024323.3 | NP_077299.3 | 5 - 6 | 452 | 95 | |
XM_006722885.3 | XP_006722948.2 | 5 - 6 | 825 | 95 | ||
XM_011528286.2 | XP_011526588.1 | 4 - 5 | 614 | 95 | ||
XM_011528287.2 | XP_011526589.1 | 4 - 5 | 274 | 95 | ||
XM_017027300.1 | XP_016882789.1 | 4 - 5 | 612 | 95 | ||
XM_017027301.1 | XP_016882790.1 | 4 - 5 | 476 | 95 | ||
GenBank mRNA | AK097431.1 | - | 5 - 6 | 452 | 95 | |
BC119719.1 | - | 4 - 5 | 420 | 40117748 | 95 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC19001225.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_011528286.2,XM_011528287.2,XM_017027300.1,XM_017027301.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000585755.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000585755.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000585755.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000586500.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000586500.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000586500.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000586500.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000588115.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000588115.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000588115.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000588115.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000588115.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000589393.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000589393.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000589393.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000589393.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000591439.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000591439.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000591439.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19017218.hg.1 | ENST00000346736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19017218.hg.1 | ENST00000454313.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19017218.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19017218.hg.1 | TC19001225.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1900180063.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900180063.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900180063.hg.1 | ENST00000589393.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900180063.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900180063.hg.1 | TC1900009791.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900180063.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900180063.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900180063.hg.1 | ENST00000589393.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900180063.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000585755.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000585755.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000585755.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000586500.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000586500.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000586500.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000586500.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000588115.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000588115.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000588115.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000588115.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000588115.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000589393.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000589393.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000589393.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000589393.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000591439.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000591439.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000591439.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | ENST00000591586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009791.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900180064.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900180064.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900180064.hg.1 | NM_024323.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900180064.hg.1 | TC1900009791.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900180064.hg.1 | ENST00000346736.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900180064.hg.1 | ENST00000586783.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900180064.hg.1 | NM_024323.4 | Refseq |
