| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03022874.hg.1 |
TC03001337.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03022874.hg.1 |
ENST00000340674.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03022874.hg.1 |
ENST00000396077.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03022874.hg.1 |
ENST00000425708.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03022874.hg.1 |
NM_181489.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
ENST00000396077.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
ENST00000396077.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
ENST00000396077.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
ENST00000396077.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
ENST00000396077.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
ENST00000396077.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
ENST00000396077.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
ENST00000396077.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
ENST00000396077.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
ENST00000396077.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
ENST00000396077.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
ENST00000396077.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
ENST00000396077.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
ENST00000396077.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
ENST00000396077.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
ENST00000396077.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
ENST00000396077.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
ENST00000396077.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
ENST00000425708.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
ENST00000425708.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
ENST00000425708.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
ENST00000425708.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
ENST00000425708.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
ENST00000425708.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
ENST00000425708.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
ENST00000425708.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
ENST00000425708.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
ENST00000425708.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
ENST00000425708.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
ENST00000425708.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
ENST00000425708.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
ENST00000425708.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
ENST00000425708.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
ENST00000425708.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
ENST00000425708.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
ENST00000460529.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
ENST00000460529.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
ENST00000460529.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
ENST00000460529.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
ENST00000460529.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
ENST00000460529.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
ENST00000460529.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
ENST00000460529.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
ENST00000474600.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
ENST00000474600.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
ENST00000474600.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
ENST00000474600.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
ENST00000474600.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
ENST00000617032.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
ENST00000617032.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
ENST00000617032.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
ENST00000617032.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
ENST00000617032.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
ENST00000617032.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
ENST00000617032.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
ENST00000617032.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
NM_181489.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
NM_181489.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
NM_181489.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
NM_181489.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
NM_181489.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
NM_181489.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
NM_181489.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
NM_181489.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
NM_181489.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
NM_181489.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
NM_181489.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
NM_181489.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
NM_181489.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
NM_181489.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
NM_181489.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
NM_181489.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
NM_181489.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
NM_181489.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010889.hg.1 |
NM_181489.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
ENST00000396077.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
ENST00000396077.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
ENST00000396077.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
ENST00000396077.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
ENST00000396077.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
ENST00000396077.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
ENST00000396077.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
ENST00000396077.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
ENST00000396077.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
ENST00000396077.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
ENST00000396077.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
ENST00000396077.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
ENST00000396077.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
ENST00000396077.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
ENST00000396077.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
ENST00000396077.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
ENST00000396077.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
ENST00000396077.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
ENST00000425708.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
ENST00000425708.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
ENST00000425708.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
ENST00000425708.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
ENST00000425708.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
ENST00000425708.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
ENST00000425708.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
ENST00000425708.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
ENST00000425708.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
ENST00000425708.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
ENST00000425708.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
ENST00000425708.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
ENST00000425708.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
ENST00000425708.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
ENST00000425708.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
ENST00000425708.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
ENST00000425708.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
ENST00000460529.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
ENST00000460529.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
ENST00000460529.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
ENST00000460529.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
ENST00000460529.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
ENST00000460529.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
ENST00000460529.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
ENST00000460529.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
ENST00000474600.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
ENST00000474600.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
ENST00000474600.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
ENST00000474600.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
ENST00000474600.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
ENST00000617032.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
ENST00000617032.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
ENST00000617032.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
ENST00000617032.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
ENST00000617032.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
ENST00000617032.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
ENST00000617032.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
ENST00000617032.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
NM_181489.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
NM_181489.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
NM_181489.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
NM_181489.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
NM_181489.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
NM_181489.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
NM_181489.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
NM_181489.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
NM_181489.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
NM_181489.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
NM_181489.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
NM_181489.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
NM_181489.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
NM_181489.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
NM_181489.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
NM_181489.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
NM_181489.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
NM_181489.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010889.hg.1 |
NM_181489.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300179408.hg.1 |
ENST00000396077.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300179408.hg.1 |
ENST00000425708.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300179408.hg.1 |
NM_181489.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300179408.hg.1 |
ENST00000396077.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300179408.hg.1 |
ENST00000425708.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300179408.hg.1 |
NM_181489.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300179408.hg.1 |
TC0300010889.hg.1 |
Clariom D TC |