Assay Details
| Gene Symbol: NAA20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016100.4 | NP_057184.1 | 2 - 3 | 187 | 77 | |
| NM_181528.3 | NP_852669.1 | 2 - 3 | 187 | 77 | ||
| GenBank mRNA | AF085355.1 | - | 2 - 3 | 150 | 77 | |
| AK311819.1 | - | 2 - 3 | 140 | 77 | ||
| AL136641.1 | - | 2 - 3 | 119 | 77 | ||
| BC005181.1 | - | 2 - 3 | 115 | 3950192 | 77 | |
| BC008446.1 | - | 2 - 3 | 136 | 4104421 | 77 | |
| BG548527.1 | - | 2 - 3 | 135 | 77 | ||
| CA867744.1 | - | 2 - 3 | 361 | 77 | ||
| CB959658.1 | - | 2 - 3 | 211 | 77 | ||
More Information
Set Membership: |
|

NAA20
Genomic Map