Assay Details
| Gene Symbol: UFD1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035247.2 | NP_001030324.2 | 6 - 7 | 624 | 141 | |
| NM_005659.6 | NP_005650.2 | 6 - 7 | 624 | 141 | ||
| XM_011530361.2 | XP_011528663.1 | 6 - 7 | 750 | 141 | ||
| GenBank mRNA | AF141201.1 | - | 5 - 6 | 494 | 141 | |
| AJ239058.1 | - | 5 - 6 | 494 | 141 | ||
| AK225877.1 | - | 6 - 7 | 581 | 141 | ||
| AK304734.1 | - | 6 - 7 | 594 | 141 | ||
| AK315998.1 | - | 6 - 7 | 575 | 141 | ||
| AY101594.1 | - | 6 - 7 | 551 | 141 | ||
| BC001049.2 | - | 6 - 7 | 571 | 3507963 | 141 | |
| BC005087.1 | - | 6 - 7 | 572 | 3830774 | 141 | |
| BM761958.1 | - | 5 - 6 | 563 | 141 | ||
| BX648483.1 | - | 6 - 7 | 556 | 141 | ||
| CR456607.1 | - | 6 - 7 | 508 | 141 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000496.hg.1
Set Membership: |
|

UFD1L
Genomic Map