Assay Details
| Gene Symbol: ATMIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015251.2 | NP_056066.2 | 1 - 2 | 357 | 163 | |
| GenBank mRNA | AK290943.1 | - | 1 - 2 | 142 | 163 | |
| BC002701.2 | - | 1 - 2 | 174 | 163 | ||
| BX375235.2 | - | 1 - 2 | 356 | 163 | ||
| CF593326.1 | - | 1 - 2 | 374 | 163 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000635.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK290943.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900008067.hg.1 | ENST00000445995.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000299575.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000299575.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000299575.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000299575.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000299575.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000299575.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000299575.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000299575.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000539819.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000539819.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000539819.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000539819.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000562969.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000562969.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000562969.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000564241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000564241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000564241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000564241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000564241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000564241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000564241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000565237.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000565237.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000565237.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000566488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000566488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000566488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000566488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000566488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000566488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | ENST00000566488.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | NM_001300728.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | NM_001300728.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | NM_001300728.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | NM_001300728.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | NM_001300728.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | NM_001300728.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | NM_001300728.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | NM_015251.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | NM_015251.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | NM_015251.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | NM_015251.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | NM_015251.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | NM_015251.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | NM_015251.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008568.hg.1 | NM_015251.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16008996.hg.1 | ENST00000299575.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16008996.hg.1 | NM_015251.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16008996.hg.1 | TC16000635.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16008999.hg.1 | ENST00000299575.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16008999.hg.1 | ENST00000539819.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16008999.hg.1 | NM_015251.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16008999.hg.1 | TC16000635.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0900008067.hg.1 | ENST00000445995.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600163952.hg.1 | ENST00000299575.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600163952.hg.1 | ENST00000562969.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600163952.hg.1 | NM_015251.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600163952.hg.1 | ENST00000299575.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600163952.hg.1 | ENST00000562969.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600163952.hg.1 | NM_015251.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600163952.hg.1 | TC1600008568.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000299575.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000299575.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000299575.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000299575.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000299575.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000299575.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000299575.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000299575.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000539819.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000539819.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000539819.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000539819.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000562969.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000562969.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000562969.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000564241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000564241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000564241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000564241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000564241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000564241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000564241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000565237.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000565237.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000565237.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000566488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000566488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000566488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000566488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000566488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000566488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | ENST00000566488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | NM_001300728.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | NM_001300728.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | NM_001300728.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | NM_001300728.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | NM_001300728.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | NM_001300728.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | NM_001300728.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | NM_015251.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | NM_015251.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | NM_015251.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | NM_015251.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | NM_015251.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | NM_015251.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | NM_015251.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008568.hg.1 | NM_015251.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600163951.hg.1 | ENST00000299575.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600163951.hg.1 | NM_015251.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600163951.hg.1 | ENST00000299575.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600163951.hg.1 | NM_015251.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600163951.hg.1 | TC1600008568.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600163958.hg.1 | ENST00000299575.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600163958.hg.1 | ENST00000539819.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600163958.hg.1 | ENST00000562969.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600163958.hg.1 | ENST00000564241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600163958.hg.1 | ENST00000565237.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600163958.hg.1 | ENST00000566488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600163958.hg.1 | NM_001300728.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600163958.hg.1 | NM_015251.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600163958.hg.1 | ENST00000299575.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600163958.hg.1 | ENST00000539819.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600163958.hg.1 | ENST00000562969.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600163958.hg.1 | ENST00000564241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600163958.hg.1 | ENST00000565237.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600163958.hg.1 | ENST00000566488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600163958.hg.1 | NM_001300728.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600163958.hg.1 | NM_015251.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600163958.hg.1 | TC1600008568.hg.1 | Clariom D TC |

ATMIN
Genomic Map