| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011664.hg.1 |
ENST00000606054.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011664.hg.1 |
ENST00000606054.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011664.hg.1 |
ENST00000606054.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011664.hg.1 |
NR_046368.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011664.hg.1 |
NR_046368.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011664.hg.1 |
NR_046368.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500180672.hg.1 |
ENST00000606054.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500180672.hg.1 |
NR_046368.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500180672.hg.1 |
ENST00000606054.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500180672.hg.1 |
NR_046368.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500180672.hg.1 |
TC0500011664.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0500011664.hg.1 |
ENST00000606054.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011664.hg.1 |
ENST00000606054.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011664.hg.1 |
ENST00000606054.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011664.hg.1 |
NR_046368.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011664.hg.1 |
NR_046368.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011664.hg.1 |
NR_046368.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500180674.hg.1 |
ENST00000606054.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500180674.hg.1 |
NR_046368.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500180674.hg.1 |
ENST00000606054.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500180674.hg.1 |
NR_046368.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500180674.hg.1 |
TC0500011664.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0500180673.hg.1 |
ENST00000606054.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500180673.hg.1 |
NR_046368.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500180673.hg.1 |
ENST00000606054.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500180673.hg.1 |
NR_046368.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500180673.hg.1 |
TC0500011664.hg.1 |
Clariom D TC |