Assay Details
| Gene Symbol: EXOC3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_126522.1 | 1 | 1060 | 87 | ||
| GenBank mRNA | BC014011.2 | - | 1 | 995 | 87 | |
| BC029796.1 | - | 1 | 1043 | 5183143 | 87 | |
| BM681006.1 | - | 1 | 87 | |||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03709710 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23873692 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23873691 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03709721 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23873693 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03709718 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23873698 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03709709 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03709723 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23873696 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15652702 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03709711 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 954235 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3217517 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P247316 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 8914 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 29614 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P31429393 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3217517 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3217517 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03709709 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03709717 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03709715 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03709720 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03709723 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23873697 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15652702 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23873693 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23873696 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03709718 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23873691 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03709718 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23873697 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03709717 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23873693 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03709715 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23873691 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03709720 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03709709 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23873696 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15652702 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03709723 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P247316 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P247316 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P15057453 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P456199 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03709720 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15652699 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03709714 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03709711 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15652706 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03709718 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NM_138464_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8110683 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8110682 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8110684 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8110681 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 235219_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11728045_at | - | - |
| Affymetrix Human Genome U219 Array | 11728045_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009801.hg.1 | ENST00000408966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009801.hg.1 | ENST00000408966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009801.hg.1 | ENST00000408966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009801.hg.1 | ENST00000623673.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009801.hg.1 | ENST00000623673.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009801.hg.1 | ENST00000623673.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009801.hg.1 | NR_126522.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009801.hg.1 | NR_126522.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500169044.hg.1 | ENST00000408966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169044.hg.1 | ENST00000623673.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169044.hg.1 | NR_126522.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500169044.hg.1 | ENST00000408966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169044.hg.1 | ENST00000623673.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169044.hg.1 | NR_126522.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500169044.hg.1 | TC0500009801.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500009801.hg.1 | ENST00000408966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009801.hg.1 | ENST00000408966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009801.hg.1 | ENST00000408966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009801.hg.1 | ENST00000623673.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009801.hg.1 | ENST00000623673.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009801.hg.1 | ENST00000623673.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009801.hg.1 | NR_126522.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009801.hg.1 | NR_126522.1 | Refseq |
| Human Genome U133 Plus 2.0 Array | 235219_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 235219_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235219_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235219_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235219_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235219_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235219_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235219_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235219_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235219_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235219_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235219_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 235219_at3 | - | - |
| Human Genome U95 Set | 83191_at | - | - |
| Human Genome U95 Set | 83191_PM_at | - | - |
| Human Genome U133 Set | 235219_at | - | - |
| Human Genome U133 Set | 235219_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A026429 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A026429 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 230763 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1796922 | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1796922 | - | - |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1796922 | - | - |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1796922 | - | - |

EXOC3-AS1
Genomic Map