Assay Details
| Gene Symbol: LINC00508 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_126452.2 | 4 | 853 | 82 | ||
| GenBank mRNA | BC032019.1 | - | 1 | 542 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000479259.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000479259.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000479259.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000479259.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000479259.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000498123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000498123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000498129.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000498129.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000498129.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000498129.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000498129.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000498129.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_006243.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_006243.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_006243.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_006243.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_006243.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_006243.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_006243.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_006243.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_006243.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_006243.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_006243.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_006243.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_006243.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_006243.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_006243.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_006243.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_006243.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011520.hg.1 | NM_006243.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012399.hg.1 | ENST00000538901.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012399.hg.1 | ENST00000538901.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012399.hg.1 | NR_126452.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012399.hg.1 | NR_126452.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012399.hg.1 | NR_126452.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012399.hg.1 | NR_126452.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200197042.hg.1 | NR_126452.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200197042.hg.1 | NR_126452.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200197042.hg.1 | TC1200012399.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200197044.hg.1 | ENST00000538901.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200197044.hg.1 | NR_126452.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200197044.hg.1 | ENST00000538901.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200197044.hg.1 | NR_126452.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200197044.hg.1 | TC1200012399.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000261461.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000479259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000479259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000479259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000479259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000479259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000498123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000498123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000498129.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000498129.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000498129.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000498129.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000498129.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000498129.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | ENST00000537030.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_001199756.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_006243.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_006243.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_006243.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_006243.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_006243.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_006243.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_006243.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_006243.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_006243.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_006243.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_006243.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_006243.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_006243.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_006243.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_006243.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_006243.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_006243.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011520.hg.1 | NM_006243.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012399.hg.1 | ENST00000538901.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012399.hg.1 | ENST00000538901.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012399.hg.1 | NR_126452.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012399.hg.1 | NR_126452.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012399.hg.1 | NR_126452.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012399.hg.1 | NR_126452.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200197046.hg.1 | NR_126452.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200197046.hg.1 | NR_126452.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200197046.hg.1 | TC1200012399.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200197047.hg.1 | NR_126452.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200197047.hg.1 | NR_126452.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200197047.hg.1 | TC1200012399.hg.1 | Clariom D TC |

LINC00508
Genomic Map