Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Gene 1.0 ST Array |
8167651 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
8167650 |
- |
- |
Affymetrix HT HG-U133+ PM Array Plate |
1555396_PM_s_at |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010139.hg.1 |
ENST00000513713.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007263.hg.1 |
ENST00000342995.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007263.hg.1 |
NM_203407.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007263.hg.1 |
ENST00000342995.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007263.hg.1 |
NM_203407.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010139.hg.1 |
ENST00000513713.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00149993.hg.1 |
ENST00000342995.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00149993.hg.1 |
NM_203407.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00149993.hg.1 |
ENST00000342995.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00149993.hg.1 |
NM_203407.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00149993.hg.1 |
TC0X00007263.hg.1 |
Clariom D TC |
Human Genome U133 Plus 2.0 Array |
1555396_s_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
1555396_PM_s_at |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1555396_s_at3 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1555396_s_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1555396_s_at9 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1555396_s_at4 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1555396_s_at6 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1555396_s_at10 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1555396_s_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1555396_s_at5 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1555396_s_at2 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1555396_s_at7 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1555396_s_at1 |
- |
- |