| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008075.hg.1 |
ENST00000392051.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008075.hg.1 |
ENST00000392051.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008075.hg.1 |
ENST00000392051.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008075.hg.1 |
ENST00000392051.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008075.hg.1 |
ENST00000392051.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008075.hg.1 |
ENST00000392051.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008075.hg.1 |
ENST00000594480.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008075.hg.1 |
ENST00000594480.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008075.hg.1 |
ENST00000594480.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008075.hg.1 |
ENST00000594480.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008075.hg.1 |
ENST00000594480.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008075.hg.1 |
ENST00000594480.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008075.hg.1 |
NM_001190441.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008075.hg.1 |
NM_001190441.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008075.hg.1 |
NM_001190441.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008075.hg.1 |
NM_001190441.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008075.hg.1 |
NM_001190441.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008075.hg.1 |
NM_001190441.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008072.hg.1 |
ENST00000221797.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008072.hg.1 |
ENST00000221797.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008072.hg.1 |
ENST00000221797.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008072.hg.1 |
ENST00000221797.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008072.hg.1 |
ENST00000221797.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008072.hg.1 |
ENST00000221797.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008072.hg.1 |
ENST00000221797.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008072.hg.1 |
ENST00000600141.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008072.hg.1 |
ENST00000600141.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008072.hg.1 |
ENST00000600141.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008072.hg.1 |
ENST00000600546.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008072.hg.1 |
ENST00000600546.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008072.hg.1 |
ENST00000600546.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008072.hg.1 |
ENST00000600546.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008072.hg.1 |
ENST00000600546.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008072.hg.1 |
ENST00000600546.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008072.hg.1 |
ENST00000621475.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008072.hg.1 |
ENST00000621475.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008072.hg.1 |
ENST00000621475.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008072.hg.1 |
ENST00000621475.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008072.hg.1 |
ENST00000621475.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008072.hg.1 |
NM_013268.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008072.hg.1 |
NM_013268.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008072.hg.1 |
NM_013268.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008072.hg.1 |
NM_013268.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008072.hg.1 |
NM_013268.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008072.hg.1 |
NM_013268.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008072.hg.1 |
NM_013268.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19007530.hg.1 |
ENST00000221797.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19007530.hg.1 |
NM_013268.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19007530.hg.1 |
TC19000551.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19007531.hg.1 |
ENST00000221797.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19007531.hg.1 |
NM_013268.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19007531.hg.1 |
TC19000551.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1900160871.hg.1 |
ENST00000221797.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900160871.hg.1 |
ENST00000600141.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900160871.hg.1 |
ENST00000621475.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900160871.hg.1 |
NM_013268.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900160871.hg.1 |
TC1900008072.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900160871.hg.1 |
ENST00000221797.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900160871.hg.1 |
ENST00000600141.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900160871.hg.1 |
ENST00000621475.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900160871.hg.1 |
NM_013268.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900160873.hg.1 |
ENST00000221797.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900160873.hg.1 |
ENST00000621475.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900160873.hg.1 |
NM_013268.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900160873.hg.1 |
TC1900008072.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900160873.hg.1 |
ENST00000221797.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900160873.hg.1 |
ENST00000621475.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900160873.hg.1 |
NM_013268.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008072.hg.1 |
ENST00000221797.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008072.hg.1 |
ENST00000221797.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008072.hg.1 |
ENST00000221797.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008072.hg.1 |
ENST00000221797.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008072.hg.1 |
ENST00000221797.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008072.hg.1 |
ENST00000221797.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008072.hg.1 |
ENST00000221797.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008072.hg.1 |
ENST00000600141.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008072.hg.1 |
ENST00000600141.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008072.hg.1 |
ENST00000600141.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008072.hg.1 |
ENST00000600546.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008072.hg.1 |
ENST00000600546.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008072.hg.1 |
ENST00000600546.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008072.hg.1 |
ENST00000600546.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008072.hg.1 |
ENST00000600546.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008072.hg.1 |
ENST00000600546.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008072.hg.1 |
ENST00000621475.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008072.hg.1 |
ENST00000621475.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008072.hg.1 |
ENST00000621475.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008072.hg.1 |
ENST00000621475.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008072.hg.1 |
ENST00000621475.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008072.hg.1 |
NM_013268.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008072.hg.1 |
NM_013268.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008072.hg.1 |
NM_013268.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008072.hg.1 |
NM_013268.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008072.hg.1 |
NM_013268.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008072.hg.1 |
NM_013268.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008072.hg.1 |
NM_013268.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008075.hg.1 |
ENST00000392051.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008075.hg.1 |
ENST00000392051.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008075.hg.1 |
ENST00000392051.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008075.hg.1 |
ENST00000392051.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008075.hg.1 |
ENST00000392051.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008075.hg.1 |
ENST00000392051.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008075.hg.1 |
ENST00000594480.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008075.hg.1 |
ENST00000594480.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008075.hg.1 |
ENST00000594480.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008075.hg.1 |
ENST00000594480.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008075.hg.1 |
ENST00000594480.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008075.hg.1 |
ENST00000594480.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008075.hg.1 |
NM_001190441.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008075.hg.1 |
NM_001190441.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008075.hg.1 |
NM_001190441.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008075.hg.1 |
NM_001190441.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008075.hg.1 |
NM_001190441.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008075.hg.1 |
NM_001190441.2 |
Refseq |