Assay Details
| Gene Symbol: GPSM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276501.1 | NP_001263430.1 | 2 - 3 | 551 | 114 | |
| NM_022107.2 | NP_071390.1 | 6 - 7 | 522 | 114 | ||
| GenBank mRNA | AF155657.1 | - | 2 - 3 | 170 | 114 | |
| AJ243937.1 | - | 6 - 7 | 522 | 114 | ||
| AJ243938.1 | - | 2 - 3 | 542 | 114 | ||
| AK313922.1 | - | 2 - 3 | 551 | 114 | ||
| BC018724.1 | - | 2 - 3 | 214 | 4576337 | 114 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06001563.hg.1, TC6_apd_hap1000097.hg.1, TC6_cox_hap2000189.hg.1, TC6_mann_hap4000154.hg.1, TC6_mcf_hap5000164.hg.1, TC6_qbl_hap6000178.hg.1, TC6_ssto_hap7000158.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2003681.hg.1 | TC6_cox_hap2000189.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2003681.hg.1 | NM_022107.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7003247.hg.1 | TC6_ssto_hap7000158.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7003247.hg.1 | NM_022107.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06018459.hg.1 | TC06001563.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06018459.hg.1 | ENST00000375040.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06018459.hg.1 | ENST00000375043.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06018459.hg.1 | ENST00000487761.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06018459.hg.1 | NM_022107.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1001554.hg.1 | TC6_apd_hap1000097.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1001554.hg.1 | NM_022107.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6003517.hg.1 | TC6_qbl_hap6000178.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6003517.hg.1 | NM_022107.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1001555.hg.1 | TC6_apd_hap1000097.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1001555.hg.1 | NM_022107.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4002819.hg.1 | TC6_mann_hap4000154.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4002819.hg.1 | NM_022107.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7003248.hg.1 | TC6_ssto_hap7000158.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7003248.hg.1 | NM_022107.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6003516.hg.1 | TC6_qbl_hap6000178.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6003516.hg.1 | NM_022107.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2003682.hg.1 | TC6_cox_hap2000189.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2003682.hg.1 | NM_022107.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06018458.hg.1 | TC06001563.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06018458.hg.1 | ENST00000375040.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06018458.hg.1 | ENST00000375043.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06018458.hg.1 | ENST00000487761.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06018458.hg.1 | NM_022107.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5003355.hg.1 | TC6_mcf_hap5000164.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5003355.hg.1 | NM_022107.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4002818.hg.1 | TC6_mann_hap4000154.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4002818.hg.1 | NM_022107.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5003356.hg.1 | TC6_mcf_hap5000164.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5003356.hg.1 | NM_022107.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014270.hg.1 | ENST00000375040.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014270.hg.1 | ENST00000375040.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014270.hg.1 | ENST00000375040.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014270.hg.1 | ENST00000375040.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014270.hg.1 | ENST00000375040.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014270.hg.1 | ENST00000375040.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014270.hg.1 | ENST00000375040.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014270.hg.1 | ENST00000375040.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014270.hg.1 | ENST00000472768.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014270.hg.1 | ENST00000472768.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014270.hg.1 | ENST00000472768.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014270.hg.1 | ENST00000472768.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014270.hg.1 | ENST00000472768.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014270.hg.1 | ENST00000472768.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014270.hg.1 | ENST00000472768.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014270.hg.1 | ENST00000487761.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014270.hg.1 | ENST00000487761.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014270.hg.1 | ENST00000487761.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014270.hg.1 | ENST00000487761.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014270.hg.1 | ENST00000487761.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014270.hg.1 | ENST00000487761.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014270.hg.1 | ENST00000487761.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014270.hg.1 | ENST00000487761.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014270.hg.1 | ENST00000619085.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014270.hg.1 | ENST00000619085.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014270.hg.1 | ENST00000619085.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014270.hg.1 | ENST00000619085.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014270.hg.1 | ENST00000619085.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014270.hg.1 | ENST00000619085.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014270.hg.1 | ENST00000619085.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014270.hg.1 | NM_001276501.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014270.hg.1 | NM_001276501.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014270.hg.1 | NM_001276501.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014270.hg.1 | NM_001276501.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014270.hg.1 | NM_001276501.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014270.hg.1 | NM_001276501.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014270.hg.1 | NM_001276501.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014270.hg.1 | NM_001276501.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014270.hg.1 | ENST00000375040.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014270.hg.1 | ENST00000375040.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014270.hg.1 | ENST00000375040.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014270.hg.1 | ENST00000375040.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014270.hg.1 | ENST00000375040.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014270.hg.1 | ENST00000375040.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014270.hg.1 | ENST00000375040.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014270.hg.1 | ENST00000375040.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014270.hg.1 | ENST00000472768.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014270.hg.1 | ENST00000472768.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014270.hg.1 | ENST00000472768.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014270.hg.1 | ENST00000472768.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014270.hg.1 | ENST00000472768.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014270.hg.1 | ENST00000472768.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014270.hg.1 | ENST00000472768.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014270.hg.1 | ENST00000487761.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014270.hg.1 | ENST00000487761.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014270.hg.1 | ENST00000487761.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014270.hg.1 | ENST00000487761.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014270.hg.1 | ENST00000487761.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014270.hg.1 | ENST00000487761.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014270.hg.1 | ENST00000487761.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014270.hg.1 | ENST00000487761.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014270.hg.1 | ENST00000619085.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014270.hg.1 | ENST00000619085.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014270.hg.1 | ENST00000619085.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014270.hg.1 | ENST00000619085.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014270.hg.1 | ENST00000619085.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014270.hg.1 | ENST00000619085.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014270.hg.1 | ENST00000619085.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014270.hg.1 | NM_001276501.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014270.hg.1 | NM_001276501.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014270.hg.1 | NM_001276501.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014270.hg.1 | NM_001276501.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014270.hg.1 | NM_001276501.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014270.hg.1 | NM_001276501.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014270.hg.1 | NM_001276501.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014270.hg.1 | NM_001276501.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600201386.hg.1 | ENST00000375040.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201386.hg.1 | ENST00000472768.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201386.hg.1 | ENST00000487761.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201386.hg.1 | ENST00000619085.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201386.hg.1 | NM_001276501.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600201386.hg.1 | ENST00000375040.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201386.hg.1 | ENST00000472768.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201386.hg.1 | ENST00000487761.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201386.hg.1 | ENST00000619085.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201386.hg.1 | NM_001276501.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600201386.hg.1 | TC0600014270.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600201387.hg.1 | ENST00000375040.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201387.hg.1 | ENST00000487761.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201387.hg.1 | ENST00000619085.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201387.hg.1 | NM_001276501.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600201387.hg.1 | ENST00000375040.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201387.hg.1 | ENST00000487761.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201387.hg.1 | ENST00000619085.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600201387.hg.1 | NM_001276501.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600201387.hg.1 | TC0600014270.hg.1 | Clariom D TC |

GPSM3
Genomic Map