Assay Details
| Gene Symbol: C1GALT1C1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152692.4 | NP_689905.1 | 2 - 3 | 405 | 99 | |
| GenBank mRNA | AY159319.1 | - | 2 - 3 | 250 | 99 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AY159319.1, NM_152692.4 transcripts.
Set Membership: |
|

C1GALT1C1
Genomic Map