Assay Details
| Gene Symbol: SLC26A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134425.2 | NP_602297.1 | 2 - 3 | 743 | 64 | |
| GenBank mRNA | BC015517.2 | - | 2 - 3 | 679 | 64 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04000959.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
ion transport
chloride transport
sulfate transport
bicarbonate transport
oxalate transport
regulation of membrane potential
3'-phosphoadenosine 5'-phosphosulfate biosynthetic process
regulation of intracellular pH
sulfate transmembrane transport
chloride transmembrane transport
chloride channel activity
secondary active sulfate transmembrane transporter activity
bicarbonate transmembrane transporter activity
chloride transmembrane transporter activity
sulfate transmembrane transporter activity
anion:anion antiporter activity
oxalate transmembrane transporter activity
ion transport
chloride transport
sulfate transport
bicarbonate transport
oxalate transport
regulation of membrane potential
3'-phosphoadenosine 5'-phosphosulfate biosynthetic process
regulation of intracellular pH
sulfate transmembrane transport
chloride transmembrane transport
chloride channel activity
secondary active sulfate transmembrane transporter activity
bicarbonate transmembrane transporter activity
chloride transmembrane transporter activity
sulfate transmembrane transporter activity
anion:anion antiporter activity
oxalate transmembrane transporter activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400009714.hg.1 | ENST00000361661.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009714.hg.1 | ENST00000361661.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009714.hg.1 | ENST00000361661.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009714.hg.1 | ENST00000361661.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009714.hg.1 | ENST00000361661.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009714.hg.1 | ENST00000361661.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009714.hg.1 | ENST00000361661.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009714.hg.1 | ENST00000361661.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009714.hg.1 | ENST00000361661.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009714.hg.1 | ENST00000398516.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009714.hg.1 | ENST00000398516.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009714.hg.1 | ENST00000398516.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009714.hg.1 | ENST00000398516.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009714.hg.1 | ENST00000398516.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009714.hg.1 | ENST00000398516.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009714.hg.1 | ENST00000398516.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009714.hg.1 | ENST00000398520.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009714.hg.1 | ENST00000398520.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009714.hg.1 | ENST00000398520.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009714.hg.1 | ENST00000398520.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009714.hg.1 | ENST00000398520.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009714.hg.1 | ENST00000398520.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009714.hg.1 | ENST00000513138.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009714.hg.1 | ENST00000513138.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009714.hg.1 | ENST00000622731.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009714.hg.1 | ENST00000622731.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009714.hg.1 | ENST00000622731.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009714.hg.1 | ENST00000622731.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009714.hg.1 | ENST00000622731.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009714.hg.1 | ENST00000622731.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009714.hg.1 | ENST00000622731.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009714.hg.1 | NM_022042.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009714.hg.1 | NM_022042.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009714.hg.1 | NM_022042.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009714.hg.1 | NM_022042.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009714.hg.1 | NM_022042.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009714.hg.1 | NM_022042.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009714.hg.1 | NM_022042.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009714.hg.1 | NM_022042.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009714.hg.1 | NM_134425.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009714.hg.1 | NM_134425.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009714.hg.1 | NM_134425.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009714.hg.1 | NM_134425.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009714.hg.1 | NM_134425.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009714.hg.1 | NM_134425.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009714.hg.1 | NM_134425.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009714.hg.1 | NM_213613.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009714.hg.1 | NM_213613.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009714.hg.1 | NM_213613.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009714.hg.1 | NM_213613.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009714.hg.1 | NM_213613.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009714.hg.1 | NM_213613.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009714.hg.1 | NM_213613.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009714.hg.1 | NM_213613.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009714.hg.1 | NM_213613.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04012967.hg.1 | TC04000959.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04012967.hg.1 | ENST00000398520.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04012967.hg.1 | NM_134425.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400166526.hg.1 | ENST00000398520.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400166526.hg.1 | ENST00000622731.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400166526.hg.1 | NM_134425.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400166526.hg.1 | ENST00000398520.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400166526.hg.1 | ENST00000622731.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400166526.hg.1 | NM_134425.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400166526.hg.1 | TC0400009714.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400166525.hg.1 | ENST00000398520.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400166525.hg.1 | NM_134425.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400166525.hg.1 | ENST00000398520.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400166525.hg.1 | NM_134425.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400166525.hg.1 | TC0400009714.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400009714.hg.1 | ENST00000361661.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009714.hg.1 | ENST00000361661.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009714.hg.1 | ENST00000361661.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009714.hg.1 | ENST00000361661.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009714.hg.1 | ENST00000361661.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009714.hg.1 | ENST00000361661.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009714.hg.1 | ENST00000361661.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009714.hg.1 | ENST00000361661.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009714.hg.1 | ENST00000361661.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009714.hg.1 | ENST00000398516.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009714.hg.1 | ENST00000398516.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009714.hg.1 | ENST00000398516.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009714.hg.1 | ENST00000398516.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009714.hg.1 | ENST00000398516.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009714.hg.1 | ENST00000398516.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009714.hg.1 | ENST00000398516.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009714.hg.1 | ENST00000398520.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009714.hg.1 | ENST00000398520.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009714.hg.1 | ENST00000398520.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009714.hg.1 | ENST00000398520.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009714.hg.1 | ENST00000398520.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009714.hg.1 | ENST00000398520.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009714.hg.1 | ENST00000513138.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009714.hg.1 | ENST00000513138.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009714.hg.1 | ENST00000622731.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009714.hg.1 | ENST00000622731.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009714.hg.1 | ENST00000622731.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009714.hg.1 | ENST00000622731.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009714.hg.1 | ENST00000622731.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009714.hg.1 | ENST00000622731.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009714.hg.1 | ENST00000622731.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009714.hg.1 | NM_022042.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009714.hg.1 | NM_022042.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009714.hg.1 | NM_022042.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009714.hg.1 | NM_022042.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009714.hg.1 | NM_022042.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009714.hg.1 | NM_022042.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009714.hg.1 | NM_022042.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009714.hg.1 | NM_022042.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009714.hg.1 | NM_134425.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009714.hg.1 | NM_134425.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009714.hg.1 | NM_134425.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009714.hg.1 | NM_134425.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009714.hg.1 | NM_134425.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009714.hg.1 | NM_134425.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009714.hg.1 | NM_134425.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009714.hg.1 | NM_213613.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009714.hg.1 | NM_213613.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009714.hg.1 | NM_213613.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009714.hg.1 | NM_213613.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009714.hg.1 | NM_213613.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009714.hg.1 | NM_213613.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009714.hg.1 | NM_213613.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009714.hg.1 | NM_213613.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009714.hg.1 | NM_213613.3 | Refseq |

SLC26A1
Genomic Map