Assay Details
| Gene Symbol: CISH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013324.5 | NP_037456.5 | 1 - 2 | 201 | 129 | |
| GenBank mRNA | AF132297.2 | - | 1 - 2 | 142 | 129 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001435.hg.1
Additional Information:
This assay is in the 5'UTR region of the AF132297.2,NM_013324.5 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cell growth
negative regulation of protein kinase activity
protein kinase C-activating G-protein coupled receptor signaling pathway
protein ubiquitination
cytokine-mediated signaling pathway
intracellular signal transduction
negative regulation of JAK-STAT cascade
negative regulation of insulin receptor signaling pathway
molecular_function
protein kinase inhibitor activity
protein binding
regulation of cell growth
negative regulation of protein kinase activity
protein kinase C-activating G-protein coupled receptor signaling pathway
protein ubiquitination
cytokine-mediated signaling pathway
intracellular signal transduction
negative regulation of JAK-STAT cascade
negative regulation of insulin receptor signaling pathway
molecular_function
protein kinase inhibitor activity
protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03025937.hg.1 | TC03001435.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03025937.hg.1 | ENST00000443053.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03025937.hg.1 | NM_013324.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03025940.hg.1 | TC03001435.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03025940.hg.1 | ENST00000348721.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03025940.hg.1 | ENST00000443053.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03025940.hg.1 | NM_013324.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03025940.hg.1 | NM_145071.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011124.hg.1 | ENST00000348721.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011124.hg.1 | ENST00000348721.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011124.hg.1 | ENST00000348721.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011124.hg.1 | ENST00000348721.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011124.hg.1 | ENST00000348721.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011124.hg.1 | ENST00000443053.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011124.hg.1 | ENST00000443053.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011124.hg.1 | ENST00000443053.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011124.hg.1 | ENST00000443053.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011124.hg.1 | ENST00000443053.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011124.hg.1 | ENST00000443053.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011124.hg.1 | ENST00000443053.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011124.hg.1 | ENST00000491847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011124.hg.1 | ENST00000491847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011124.hg.1 | ENST00000491847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011124.hg.1 | ENST00000491847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011124.hg.1 | ENST00000491847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011124.hg.1 | ENST00000491847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011124.hg.1 | ENST00000491847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011124.hg.1 | ENST00000491847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011124.hg.1 | ENST00000491847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011124.hg.1 | NM_013324.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011124.hg.1 | NM_013324.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011124.hg.1 | NM_013324.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011124.hg.1 | NM_013324.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011124.hg.1 | NM_013324.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011124.hg.1 | NM_013324.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011124.hg.1 | NM_013324.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011124.hg.1 | NM_013324.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011124.hg.1 | NM_013324.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011124.hg.1 | NM_145071.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011124.hg.1 | NM_145071.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011124.hg.1 | NM_145071.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011124.hg.1 | NM_145071.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011124.hg.1 | NM_145071.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011124.hg.1 | NM_145071.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011124.hg.1 | NM_145071.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03025939.hg.1 | TC03001435.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03025939.hg.1 | ENST00000348721.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03025939.hg.1 | ENST00000443053.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03025939.hg.1 | NM_013324.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03025939.hg.1 | NM_145071.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03025943.hg.1 | TC03001435.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03025943.hg.1 | ENST00000348721.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03025943.hg.1 | NM_013324.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03025943.hg.1 | NM_145071.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300182936.hg.1 | ENST00000348721.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300182936.hg.1 | NM_013324.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300182936.hg.1 | NM_145071.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300182936.hg.1 | ENST00000348721.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300182936.hg.1 | NM_013324.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300182936.hg.1 | NM_145071.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300182936.hg.1 | TC0300011124.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300182933.hg.1 | ENST00000348721.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300182933.hg.1 | ENST00000443053.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300182933.hg.1 | ENST00000491847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300182933.hg.1 | NM_013324.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300182933.hg.1 | NM_145071.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300182933.hg.1 | ENST00000348721.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300182933.hg.1 | ENST00000443053.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300182933.hg.1 | ENST00000491847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300182933.hg.1 | NM_013324.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300182933.hg.1 | NM_145071.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300182933.hg.1 | TC0300011124.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300011124.hg.1 | ENST00000348721.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011124.hg.1 | ENST00000348721.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011124.hg.1 | ENST00000348721.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011124.hg.1 | ENST00000348721.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011124.hg.1 | ENST00000348721.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011124.hg.1 | ENST00000443053.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011124.hg.1 | ENST00000443053.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011124.hg.1 | ENST00000443053.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011124.hg.1 | ENST00000443053.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011124.hg.1 | ENST00000443053.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011124.hg.1 | ENST00000443053.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011124.hg.1 | ENST00000443053.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011124.hg.1 | ENST00000491847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011124.hg.1 | ENST00000491847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011124.hg.1 | ENST00000491847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011124.hg.1 | ENST00000491847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011124.hg.1 | ENST00000491847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011124.hg.1 | ENST00000491847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011124.hg.1 | ENST00000491847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011124.hg.1 | ENST00000491847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011124.hg.1 | ENST00000491847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011124.hg.1 | NM_013324.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011124.hg.1 | NM_013324.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011124.hg.1 | NM_013324.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011124.hg.1 | NM_013324.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011124.hg.1 | NM_013324.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011124.hg.1 | NM_013324.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011124.hg.1 | NM_013324.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011124.hg.1 | NM_013324.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011124.hg.1 | NM_013324.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011124.hg.1 | NM_145071.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011124.hg.1 | NM_145071.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011124.hg.1 | NM_145071.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011124.hg.1 | NM_145071.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011124.hg.1 | NM_145071.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011124.hg.1 | NM_145071.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011124.hg.1 | NM_145071.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300182929.hg.1 | ENST00000443053.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300182929.hg.1 | ENST00000491847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300182929.hg.1 | NM_013324.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300182929.hg.1 | ENST00000443053.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300182929.hg.1 | ENST00000491847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300182929.hg.1 | NM_013324.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300182929.hg.1 | TC0300011124.hg.1 | Clariom D TC |

CISH
Genomic Map