Assay Details
| Gene Symbol: SUMO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006936.2 | NP_008867.2 | 2 - 3 | 313 | 155 | |
| GenBank mRNA | AK222591.1 | - | 2 - 3 | 244 | 155 | |
| AK312350.1 | - | 2 - 3 | 244 | 155 | ||
| BC000036.2 | - | 2 - 3 | 238 | 3505840 | 155 | |
| BC008420.1 | - | 2 - 3 | 313 | 4091864 | 155 | |
| BT007008.1 | - | 2 - 3 | 152 | 155 | ||
| CR542173.1 | - | 2 - 3 | 152 | 155 | ||
| CR542188.1 | - | 2 - 3 | 152 | 155 | ||
| X99584.1 | - | 2 - 3 | 246 | 155 | ||
More Information
Set Membership: |
|

SUMO3
Genomic Map