Assay Details
| Gene Symbol: SCARA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016240.2 | NP_057324.2 | 5 | 1366 | 129 | |
| NM_182826.1 | NP_878185.1 | 5 | 1366 | 129 | ||
| XM_017013535.1 | XP_016869024.1 | 6 | 1413 | 129 | ||
| XM_017013536.1 | XP_016869025.1 | 5 | 1537 | 129 | ||
| XM_017013537.1 | XP_016869026.1 | 5 | 1537 | 129 | ||
| GenBank mRNA | AB007829.1 | - | 5 | 1366 | 129 | |
| AB007830.1 | - | 5 | 1366 | 129 | ||
| BC060811.1 | - | 5 | 1558 | 30335021 | 129 | |
| BC075067.3 | - | 5 | 1245 | 30915376 | 129 | |
| BC075068.2 | - | 5 | 1245 | 30915589 | 129 | |
More Information
Updated Mapping Information
Set Membership: |
|

SCARA3
Genomic Map