Assay Details
| Gene Symbol: PSG7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290042.1 | NP_001276971.1 | 1 - 2 | 164 | 101 | |
| NM_002783.2 | NP_002774.2 | 1 - 2 | 164 | 101 | ||
| GenBank mRNA | BC030979.1 | - | 1 - 2 | 159 | 101 | |
| BG434892.1 | - | 1 - 2 | 160 | 101 | ||
| BP321474.1 | - | 1 - 2 | 163 | 101 | ||
| BX952498.1 | - | 1 - 2 | 162 | 101 | ||
| U18467.1 | - | 1 - 2 | 114 | 101 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001290042.1 transcripts.
Set Membership: |
|

PSG7
Genomic Map