Assay Details
| Gene Symbol: KRT7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005556.3 | NP_005547.3 | 7 - 8 | 1332 | 125 | |
| XM_017019294.1 | XP_016874783.1 | 7 - 8 | 1058 | 125 | ||
| GenBank mRNA | AF509887.1 | - | 7 - 8 | 1261 | 125 | |
| AJ238246.1 | - | 1720 | 125 | |||
| AK025881.1 | - | 7 - 8 | 1259 | 125 | ||
| AK128505.1 | - | 6 - 7 | 4000 | 125 | ||
| AK128848.1 | - | 8 - 9 | 1187 | 125 | ||
| AK129924.1 | - | 8 - 9 | 1266 | 125 | ||
| AK225498.1 | - | 7 - 8 | 1255 | 125 | ||
| BC002700.2 | - | 7 - 8 | 1247 | 3610347 | 125 | |
| BC009521.1 | - | 3 - 4 | 419 | 125 | ||
| BE740515.1 | - | 3 - 4 | 463 | 125 | ||
| X03212.1 | - | 7 - 8 | 1266 | 125 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000418.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000546856.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000546856.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000546856.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000546856.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000546856.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000546856.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000546856.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000547613.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000547613.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000547613.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000547613.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000547613.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000547613.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000547613.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000548088.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000548088.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000548088.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000548088.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000548088.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000548657.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000548657.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000548657.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000548657.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000549127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000549127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000549127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000549127.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000549638.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000549638.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000549638.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000549638.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000549638.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000549638.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000550153.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000550153.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000550153.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000550153.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000550153.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000550153.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000551130.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000551130.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000551130.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000551130.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000551130.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000552183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000552183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000552183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000552183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000552322.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000552322.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000552322.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000552322.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000552322.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000552322.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000552322.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000552322.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000552322.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | ENST00000552322.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12005354.hg.1 | ENST00000331817.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12005354.hg.1 | ENST00000422319.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12005354.hg.1 | ENST00000552322.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12005354.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12005354.hg.1 | TC12000418.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000546856.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000546856.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000546856.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000546856.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000546856.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000546856.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000546856.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000547613.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000547613.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000547613.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000547613.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000547613.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000547613.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000547613.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000548088.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000548088.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000548088.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000548088.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000548088.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000548657.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000548657.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000548657.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000548657.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000549127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000549127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000549127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000549127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000549638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000549638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000549638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000549638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000549638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000549638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000550153.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000550153.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000550153.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000550153.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000550153.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000550153.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000551130.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000551130.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000551130.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000551130.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000551130.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000552183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000552183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000552183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000552183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000552322.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000552322.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000552322.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000552322.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000552322.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000552322.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000552322.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000552322.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000552322.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | ENST00000552322.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012634.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200200061.hg.1 | ENST00000548657.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200200061.hg.1 | ENST00000549127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200200061.hg.1 | ENST00000549638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200200061.hg.1 | ENST00000550153.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200200061.hg.1 | ENST00000552322.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200200061.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200200061.hg.1 | ENST00000548657.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200200061.hg.1 | ENST00000549127.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200200061.hg.1 | ENST00000549638.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200200061.hg.1 | ENST00000550153.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200200061.hg.1 | ENST00000552322.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200200061.hg.1 | NM_005556.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200200061.hg.1 | TC1200012634.hg.1 | Clariom D TC |

KRT7
Genomic Map