Assay Details
| Gene Symbol: TRIM63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032588.3 | NP_115977.2 | 6 - 7 | 998 | 94 | |
| XM_017002559.1 | XP_016858048.1 | 6 - 7 | 998 | 94 | ||
| GenBank mRNA | AF353673.1 | - | 6 - 7 | 977 | 94 | |
| AJ276484.1 | - | 6 - 7 | 981 | 94 | ||
| AK056942.1 | - | 6 - 7 | 994 | 94 | ||
| AK297820.1 | - | 6 - 7 | 910 | 94 | ||
| BC080529.1 | - | 6 - 7 | 976 | 4862321 | 94 | |
| BT007373.1 | - | 6 - 7 | 858 | 94 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002386.hg.1
Set Membership: |
|

TRIM63
Genomic Map