Assay Details
| Gene Symbol: RPL21P44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027153.1 | 1 | 1268 | 92 | ||
| GenBank mRNA | BC057822.1 | - | 2 | 1239 | 92 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 73001 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 224906 | AK057097.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 224906 | AK309516.1 | genbank_mrna_human |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P94087 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 9988 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 1122 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 1191 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P00975318 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P94087 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P118258 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_32_P94087 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23257823 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03103505 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23257840 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23257799 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23257814 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03103506 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23257817 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23257845 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03103510 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03103517 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23257815 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23257808 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23257816 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23257810 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23257847 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23257835 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03103488 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23257830 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23257822 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23257797 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23257794 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03103520 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23257828 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23257821 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23257819 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23257842 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03103489 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03103486 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03103518 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23257841 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23257796 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03103490 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23257818 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03103483 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23257834 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15567656 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23257836 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03103511 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23257824 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03103482 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03103511 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03103486 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03103510 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23257817 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23257835 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23257821 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23257815 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23257840 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23257808 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23257842 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03103482 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23257822 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23257819 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03103520 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23257799 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03103488 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23257836 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23257818 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15567656 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03103505 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03103506 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23257830 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23257823 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23257834 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23257796 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03103518 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03103489 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23257828 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23257794 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23257797 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03103483 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03103490 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03103517 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23257810 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23257814 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23257845 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23257816 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23257824 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23257847 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23257841 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_32_P94087 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P41629 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03103467 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03103505 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03103488 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03103532 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03103477 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03103535 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03103528 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03103481 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03103476 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03103503 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03103460 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03103497 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03103524 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15567652 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03103526 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03103517 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03103471 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03103521 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15567651 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03103464 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03103499 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03103474 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03103538 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03103490 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03103492 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03103485 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03103494 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03103483 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03103465 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03103515 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03103510 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03103469 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03103501 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03103508 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P00975318 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 8100380 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8100381 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8100379 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8100378 | - | - |
| Human Genome U133 Plus 2.0 Array | 216479_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 216479_at | - | - |
| Human Genome U133A 2.0 Array | 216479_at | - | - |
| Human Genome U133A 2.0 Array | 216479_PM_at | - | - |
| Human Genome U133 Set | 216479_PM_at | - | - |
| Human Genome U133 Set | 216479_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A024186 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A024185 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A024185 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A024186 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 115606 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1723909 | BC057822.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1723909 | NR_027153.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1723909 | BC057822.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1723909 | NR_027153.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1723909 | BC057822.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1723909 | NR_027153.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1723909 | BC057822.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1723909 | NR_027153.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1723909 | BC057822.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1723909 | NR_027153.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1723909 | BC057822.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1723909 | NR_027153.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1723909 | BC057822.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1723909 | NR_027153.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1723909 | BC057822.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1723909 | NR_027153.1 | human_transcriptome |

RPL21P44
Genomic Map