Assay Details
| Gene Symbol: CCL3L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_111964.1 | 1 | 268 | 148 | ||
| GenBank mRNA | X52149.1 | - | 1 | 254 | 148 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
monocyte chemotaxis
inflammatory response
G-protein coupled receptor signaling pathway
negative regulation of cell proliferation
neutrophil chemotaxis
positive regulation of GTPase activity
lymphocyte chemotaxis
positive regulation of inflammatory response
chemokine-mediated signaling pathway
positive regulation of ERK1 and ERK2 cascade
cellular response to interferon-gamma
cellular response to interleukin-1
cellular response to tumor necrosis factor
chemokine activity
CCR chemokine receptor binding
monocyte chemotaxis
inflammatory response
G-protein coupled receptor signaling pathway
negative regulation of cell proliferation
neutrophil chemotaxis
positive regulation of GTPase activity
lymphocyte chemotaxis
positive regulation of inflammatory response
chemokine-mediated signaling pathway
positive regulation of ERK1 and ERK2 cascade
cellular response to interferon-gamma
cellular response to interleukin-1
cellular response to tumor necrosis factor
chemokine activity
CCR chemokine receptor binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700007618.hg.1 | ENST00000441575.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007618.hg.1 | ENST00000441575.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007618.hg.1 | ENST00000588864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007618.hg.1 | ENST00000588864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007618.hg.1 | ENST00000588864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007618.hg.1 | ENST00000590992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007618.hg.1 | ENST00000590992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007618.hg.1 | ENST00000590992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007618.hg.1 | ENST00000590992.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007618.hg.1 | ENST00000592728.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007618.hg.1 | ENST00000592728.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007618.hg.1 | ENST00000592728.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007618.hg.1 | ENST00000592728.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007618.hg.1 | ENST00000610845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007618.hg.1 | ENST00000610845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007618.hg.1 | ENST00000610845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007618.hg.1 | ENST00000615750.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007618.hg.1 | ENST00000615750.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007618.hg.1 | ENST00000616926.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007618.hg.1 | ENST00000616926.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007618.hg.1 | ENST00000616926.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007618.hg.1 | ENST00000616926.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007618.hg.1 | ENST00000620056.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007618.hg.1 | ENST00000620056.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007618.hg.1 | ENST00000620056.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007618.hg.1 | ENST00000620056.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010461.hg.1 | ENST00000612839.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010461.hg.1 | ENST00000612839.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010461.hg.1 | ENST00000612839.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010461.hg.1 | ENST00000612839.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010461.hg.1 | ENST00000612839.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010461.hg.1 | ENST00000612839.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010461.hg.1 | ENST00000612839.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010461.hg.1 | ENST00000612839.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010461.hg.1 | ENST00000612839.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010461.hg.1 | ENST00000612839.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010461.hg.1 | ENST00000612839.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010461.hg.1 | ENST00000616702.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010461.hg.1 | ENST00000616702.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010461.hg.1 | ENST00000616702.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010461.hg.1 | ENST00000616702.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010461.hg.1 | ENST00000616702.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010461.hg.1 | ENST00000619989.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010461.hg.1 | ENST00000619989.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010461.hg.1 | ENST00000619989.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010461.hg.1 | ENST00000619989.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010461.hg.1 | ENST00000619989.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010461.hg.1 | ENST00000619989.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010461.hg.1 | ENST00000619989.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010461.hg.1 | ENST00000619989.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010461.hg.1 | NM_001001437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010461.hg.1 | NM_001001437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010461.hg.1 | NM_001001437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010461.hg.1 | NM_001001437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010461.hg.1 | NM_001001437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010461.hg.1 | NM_001001437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010461.hg.1 | NM_001001437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010461.hg.1 | NM_001001437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007618.hg.1 | ENST00000441575.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007618.hg.1 | ENST00000441575.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007618.hg.1 | ENST00000588864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007618.hg.1 | ENST00000588864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007618.hg.1 | ENST00000588864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007618.hg.1 | ENST00000590992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007618.hg.1 | ENST00000590992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007618.hg.1 | ENST00000590992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007618.hg.1 | ENST00000590992.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007618.hg.1 | ENST00000592728.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007618.hg.1 | ENST00000592728.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007618.hg.1 | ENST00000592728.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007618.hg.1 | ENST00000592728.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007618.hg.1 | ENST00000610845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007618.hg.1 | ENST00000610845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007618.hg.1 | ENST00000610845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007618.hg.1 | ENST00000615750.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007618.hg.1 | ENST00000615750.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007618.hg.1 | ENST00000616926.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007618.hg.1 | ENST00000616926.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007618.hg.1 | ENST00000616926.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007618.hg.1 | ENST00000616926.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007618.hg.1 | ENST00000620056.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007618.hg.1 | ENST00000620056.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007618.hg.1 | ENST00000620056.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007618.hg.1 | ENST00000620056.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010461.hg.1 | ENST00000612839.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010461.hg.1 | ENST00000612839.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010461.hg.1 | ENST00000612839.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010461.hg.1 | ENST00000612839.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010461.hg.1 | ENST00000612839.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010461.hg.1 | ENST00000612839.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010461.hg.1 | ENST00000612839.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010461.hg.1 | ENST00000612839.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010461.hg.1 | ENST00000612839.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010461.hg.1 | ENST00000612839.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010461.hg.1 | ENST00000612839.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010461.hg.1 | ENST00000616702.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010461.hg.1 | ENST00000616702.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010461.hg.1 | ENST00000616702.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010461.hg.1 | ENST00000616702.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010461.hg.1 | ENST00000616702.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010461.hg.1 | ENST00000619989.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010461.hg.1 | ENST00000619989.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010461.hg.1 | ENST00000619989.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010461.hg.1 | ENST00000619989.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010461.hg.1 | ENST00000619989.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010461.hg.1 | ENST00000619989.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010461.hg.1 | ENST00000619989.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010461.hg.1 | ENST00000619989.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010461.hg.1 | NM_001001437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010461.hg.1 | NM_001001437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010461.hg.1 | NM_001001437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010461.hg.1 | NM_001001437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010461.hg.1 | NM_001001437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010461.hg.1 | NM_001001437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010461.hg.1 | NM_001001437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010461.hg.1 | NM_001001437.3 | Refseq |

CCL3L1
Genomic Map