Assay Details
| Gene Symbol: NT5C3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002009.2 | NP_001002009.1 | 2 - 3 | 289 | 123 | |
| NM_016489.12 | NP_057573.2 | 2 - 3 | 353 | 123 | ||
| GenBank mRNA | BC015856.1 | - | 2 - 3 | 98 | 4669009 | 123 |
| BC066914.1 | - | 2 - 3 | 214 | 5264738 | 123 | |
| BF790086.1 | - | 2 - 3 | 99 | 123 | ||
| CB137440.1 | - | 2 - 3 | 286 | 123 | ||
| DA975244.1 | - | 2 - 3 | 353 | 123 | ||
More Information
Updated Mapping Information
Set Membership: |
|

NT5C3A
Genomic Map