Assay Details
| Gene Symbol: GSTO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191014.1 | NP_001177943.1 | 3 - 4 | 528 | 139 | |
| NM_183239.1 | NP_899062.1 | 5 - 6 | 1093 | 139 | ||
| XM_011539270.2 | XP_011537572.1 | 5 - 6 | 2156 | 139 | ||
| GenBank mRNA | AK291886.1 | - | 5 - 6 | 781 | 139 | |
| AY191318.1 | - | 4 - 5 | 461 | 139 | ||
| AY209189.1 | - | 5 - 6 | 785 | 139 | ||
| AY350731.1 | - | 5 - 6 | 785 | 139 | ||
| BC023583.2 | - | 3 - 4 | 494 | 139 | ||
| BC056918.1 | - | 5 - 6 | 1093 | 5165084 | 139 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000782.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
glutathione metabolic process
xenobiotic metabolic process
L-ascorbic acid metabolic process
oxidation-reduction process
cellular response to arsenic-containing substance
cellular oxidant detoxification
glutathione derivative biosynthetic process
glutathione transferase activity
protein binding
oxidoreductase activity
glutathione dehydrogenase (ascorbate) activity
methylarsonate reductase activity
glutathione metabolic process
xenobiotic metabolic process
L-ascorbic acid metabolic process
oxidation-reduction process
cellular response to arsenic-containing substance
cellular oxidant detoxification
glutathione derivative biosynthetic process
glutathione transferase activity
protein binding
oxidoreductase activity
glutathione dehydrogenase (ascorbate) activity
methylarsonate reductase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000338595.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000338595.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000338595.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000338595.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000338595.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000338595.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000338595.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000338595.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000338595.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000338595.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000450629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000450629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000450629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000450629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000450629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000450629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000450629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000450629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000450629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000450629.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000467629.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000467629.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000467629.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000467629.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000467629.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000473401.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000473401.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000473401.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000473401.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000473401.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000477078.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000477078.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000477078.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000477078.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000477078.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000477078.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000477078.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000498052.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000498052.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000498052.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | ENST00000498052.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_001191013.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_001191013.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_001191013.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_001191013.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_001191013.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_001191013.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_001191013.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_001191013.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_001191013.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_001191013.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_001191014.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_001191014.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_001191014.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_001191014.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_001191014.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_001191014.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_001191014.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_001191014.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_001191015.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_001191015.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_001191015.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_001191015.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_001191015.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_001191015.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_001191015.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_183239.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_183239.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_183239.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_183239.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_183239.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_183239.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_183239.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_183239.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_183239.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_183239.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012498.hg.1 | NM_183239.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009344.hg.1 | ENST00000338595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009344.hg.1 | ENST00000369707.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009344.hg.1 | ENST00000369708.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009344.hg.1 | ENST00000401888.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009344.hg.1 | NM_001191014.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009344.hg.1 | NM_183239.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009344.hg.1 | TC10000782.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1000186270.hg.1 | ENST00000338595.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186270.hg.1 | ENST00000477078.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186270.hg.1 | NM_001191014.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186270.hg.1 | NM_183239.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186270.hg.1 | ENST00000338595.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186270.hg.1 | ENST00000477078.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000186270.hg.1 | NM_001191014.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186270.hg.1 | NM_183239.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000186270.hg.1 | TC1000012498.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000338595.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000338595.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000338595.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000338595.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000338595.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000338595.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000338595.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000338595.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000338595.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000338595.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000450629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000450629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000450629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000450629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000450629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000450629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000450629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000450629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000450629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000450629.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000467629.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000467629.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000467629.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000467629.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000467629.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000473401.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000473401.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000473401.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000473401.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000473401.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000477078.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000477078.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000477078.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000477078.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000477078.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000477078.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000477078.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000498052.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000498052.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000498052.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | ENST00000498052.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_001191013.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_001191013.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_001191013.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_001191013.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_001191013.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_001191013.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_001191013.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_001191013.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_001191013.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_001191013.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_001191014.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_001191014.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_001191014.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_001191014.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_001191014.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_001191014.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_001191014.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_001191014.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_001191015.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_001191015.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_001191015.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_001191015.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_001191015.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_001191015.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_001191015.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_183239.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_183239.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_183239.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_183239.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_183239.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_183239.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_183239.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_183239.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_183239.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_183239.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012498.hg.1 | NM_183239.1 | Refseq |

GSTO2
Genomic Map