Assay Details
| Gene Symbol: ZPBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198844.2 | NP_942141.2 | 1 - 2 | 203 | 143 | |
| GenBank mRNA | AY251604.1 | - | 1 - 2 | 186 | 143 | |
| BC043152.2 | - | 1 - 2 | 189 | 5297840 | 143 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000482.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000348931.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000348931.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000348931.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000348931.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000348931.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000348931.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000348931.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000348931.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000348931.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000348931.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000348931.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000377940.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000377940.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000377940.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000377940.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000377940.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000377940.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000377940.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000377940.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000377940.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000377940.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000583811.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000583811.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000583811.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000583811.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000583811.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000583811.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000584588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000584588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000584588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000584588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000584588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000584588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000584588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000584588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | ENST00000584588.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | NM_198844.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | NM_198844.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | NM_198844.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | NM_198844.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | NM_198844.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | NM_198844.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | NM_198844.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | NM_198844.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | NM_198844.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | NM_198844.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | NM_199321.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | NM_199321.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | NM_199321.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | NM_199321.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | NM_199321.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | NM_199321.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | NM_199321.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | NM_199321.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | NM_199321.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | NM_199321.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007761.hg.1 | NM_199321.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17005743.hg.1 | ENST00000348931.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17005743.hg.1 | ENST00000377940.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17005743.hg.1 | NM_198844.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17005743.hg.1 | NM_199321.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17005743.hg.1 | TC17000482.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000348931.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000348931.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000348931.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000348931.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000348931.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000348931.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000348931.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000348931.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000348931.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000348931.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000348931.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000377940.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000377940.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000377940.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000377940.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000377940.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000377940.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000377940.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000377940.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000377940.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000377940.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000583811.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000583811.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000583811.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000583811.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000583811.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000583811.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000584588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000584588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000584588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000584588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000584588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000584588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000584588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000584588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | ENST00000584588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | NM_198844.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | NM_198844.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | NM_198844.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | NM_198844.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | NM_198844.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | NM_198844.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | NM_198844.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | NM_198844.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | NM_198844.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | NM_198844.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | NM_199321.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | NM_199321.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | NM_199321.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | NM_199321.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | NM_199321.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | NM_199321.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | NM_199321.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | NM_199321.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | NM_199321.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | NM_199321.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007761.hg.1 | NM_199321.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700156308.hg.1 | ENST00000348931.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700156308.hg.1 | ENST00000377940.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700156308.hg.1 | ENST00000583811.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700156308.hg.1 | ENST00000584588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700156308.hg.1 | NM_198844.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700156308.hg.1 | NM_199321.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700156308.hg.1 | ENST00000348931.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700156308.hg.1 | ENST00000377940.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700156308.hg.1 | ENST00000583811.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700156308.hg.1 | ENST00000584588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700156308.hg.1 | NM_198844.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700156308.hg.1 | NM_199321.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700156308.hg.1 | TC1700007761.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700156309.hg.1 | ENST00000348931.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700156309.hg.1 | ENST00000377940.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700156309.hg.1 | ENST00000583811.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700156309.hg.1 | ENST00000584588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700156309.hg.1 | NM_198844.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700156309.hg.1 | NM_199321.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700156309.hg.1 | ENST00000348931.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700156309.hg.1 | ENST00000377940.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700156309.hg.1 | ENST00000583811.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700156309.hg.1 | ENST00000584588.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700156309.hg.1 | NM_198844.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700156309.hg.1 | NM_199321.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700156309.hg.1 | TC1700007761.hg.1 | Clariom D TC |

ZPBP2
Genomic Map