Assay Details
| Gene Symbol: LYPD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195685.1 | NP_001182614.1 | 2 - 3 | 412 | 132 | |
| NM_194317.3 | NP_919298.1 | 2 - 3 | 402 | 132 | ||
| GenBank mRNA | AK124733.1 | - | 2 - 3 | 412 | 132 | |
| AK309248.1 | - | 2 - 3 | 254 | 132 | ||
| AY358623.1 | - | 2 - 3 | 318 | 132 | ||
| BC032836.1 | - | 4 - 5 | 622 | 132 | ||
| BC047013.1 | - | 3 - 4 | 432 | 5171270 | 132 | |
| DA246384.1 | - | 2 - 3 | 403 | 132 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000929.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000334166.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000334166.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000334166.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000334166.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000334166.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000334166.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000334166.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000334166.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000334166.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000334166.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000334166.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000334166.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000409381.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000409381.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000409381.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000409381.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000409381.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000409381.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000409381.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000409381.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000409381.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000409381.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000414420.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000414420.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000414420.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000414420.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000414420.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000414420.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000418762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000418762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000418762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000418762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000418762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000418762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000418762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000418762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000418762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | ENST00000418762.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | NM_001195685.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | NM_001195685.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | NM_001195685.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | NM_001195685.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | NM_001195685.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | NM_001195685.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | NM_001195685.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | NM_001195685.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | NM_001195685.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | NM_001195685.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | NM_001195685.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009626.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02013617.hg.1 | TC02000929.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02013617.hg.1 | ENST00000334166.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02013617.hg.1 | ENST00000409381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02013617.hg.1 | NM_001195685.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02013617.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02013605.hg.1 | TC02000929.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02013605.hg.1 | ENST00000409381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02013605.hg.1 | NM_001195685.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200167330.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167330.hg.1 | ENST00000414420.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167330.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167330.hg.1 | ENST00000414420.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167330.hg.1 | TC0200009626.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200167334.hg.1 | ENST00000334166.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167334.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167334.hg.1 | ENST00000409381.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167334.hg.1 | ENST00000414420.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167334.hg.1 | ENST00000418762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167334.hg.1 | NM_001195685.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200167334.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200167334.hg.1 | ENST00000334166.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167334.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167334.hg.1 | ENST00000409381.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167334.hg.1 | ENST00000414420.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167334.hg.1 | ENST00000418762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167334.hg.1 | NM_001195685.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200167334.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200167334.hg.1 | TC0200009626.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000334166.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000334166.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000334166.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000334166.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000334166.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000334166.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000334166.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000334166.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000334166.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000334166.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000334166.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000334166.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000409381.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000409381.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000409381.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000409381.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000409381.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000409381.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000409381.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000409381.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000409381.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000409381.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000414420.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000414420.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000414420.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000414420.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000414420.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000414420.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000418762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000418762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000418762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000418762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000418762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000418762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000418762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000418762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000418762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | ENST00000418762.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | NM_001195685.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | NM_001195685.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | NM_001195685.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | NM_001195685.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | NM_001195685.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | NM_001195685.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | NM_001195685.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | NM_001195685.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | NM_001195685.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | NM_001195685.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | NM_001195685.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009626.hg.1 | NM_194317.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200167323.hg.1 | ENST00000409381.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167323.hg.1 | NM_001195685.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200167323.hg.1 | ENST00000409381.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167323.hg.1 | NM_001195685.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200167323.hg.1 | TC0200009626.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200167331.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167331.hg.1 | ENST00000392854.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167331.hg.1 | TC0200009626.hg.1 | Clariom D TC |

LYPD6
Genomic Map