Assay Details
| Gene Symbol: BRCC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001018055.2 | NP_001018065.1 | 5 - 6 | 516 | 143 | |
| NM_001242640.1 | NP_001229569.1 | 5 - 6 | 519 | 143 | ||
| NM_024332.3 | NP_077308.1 | 5 - 6 | 516 | 143 | ||
| XM_005274751.4 | XP_005274808.1 | 5 - 6 | 529 | 143 | ||
| XM_017029838.1 | XP_016885327.1 | 5 - 6 | 533 | 143 | ||
| GenBank mRNA | AK298886.1 | - | 4 - 5 | 351 | 143 | |
| AK299194.1 | - | 5 - 6 | 414 | 143 | ||
| AK310286.1 | - | 5 - 6 | 417 | 143 | ||
| AK313544.1 | - | 5 - 6 | 411 | 143 | ||
| AY438030.1 | - | 5 - 6 | 408 | 143 | ||
| BC002999.1 | - | 5 - 6 | 410 | 2821917 | 143 | |
| BC006540.2 | - | 5 - 6 | 410 | 2821917 | 143 | |
| X64643.1 | - | 5 - 6 | 414 | 143 | ||
More Information
Set Membership: |
|

BRCC3
Genomic Map