Assay Details
| Gene Symbol: EXOC3L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC034287.1 | - | 1 - 2 | 469 | 146 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000562887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000562887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000562887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000562887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000562887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000562887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000563536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000563536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000563536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000563536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000563536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000564324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000564324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000564324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000564324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000564324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000564324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000564324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000564324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000564324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000564324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000564324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000564324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000564324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000564324.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000564418.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000564418.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000564418.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000564418.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000564418.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000564418.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000564418.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | ENST00000564418.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000314586.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000545725.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000562887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000562887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000562887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000562887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000562887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000562887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000563536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000563536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000563536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000563536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000563536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000563889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000564324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000564324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000564324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000564324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000564324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000564324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000564324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000564324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000564324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000564324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000564324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000564324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000564324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000564324.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000564418.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000564418.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000564418.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000564418.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000564418.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000564418.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000564418.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | ENST00000564418.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | NM_178516.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010618.hg.1 | NM_178516.3 | Refseq |

EXOC3L1
Genomic Map