Assay Details
| Gene Symbol: TXN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244938.1 | NP_001231867.1 | 1 - 2 | 223 | 151 | |
| NM_003329.3 | NP_003320.2 | 1 - 2 | 223 | 151 | ||
| GenBank mRNA | AK289508.1 | - | 1 - 2 | 144 | 151 | |
| AW160756.1 | - | 1 - 2 | 223 | 151 | ||
| BC054866.1 | - | 1 - 2 | 94 | 6463280 | 151 | |
| BX647247.1 | - | 1 - 2 | 124 | 151 | ||
| X77584.1 | - | 1 - 2 | 98 | 151 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09001473.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK289508.1,BC054866.1,NM_001244938.1,NM_003329.3,X77584.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
sulfate assimilation
negative regulation of transcription from RNA polymerase II promoter
response to reactive oxygen species
transcription, DNA-templated
protein folding
glycerol ether metabolic process
movement of cell or subcellular component
signal transduction
cell-cell signaling
cell proliferation
response to radiation
nucleobase-containing small molecule interconversion
protein repair
activation of protein kinase B activity
positive regulation of peptidyl-serine phosphorylation
regulation of protein import into nucleus, translocation
positive regulation of DNA binding
cell redox homeostasis
negative regulation of protein export from nucleus
positive regulation of protein kinase B signaling
oxidation-reduction process
negative regulation of hydrogen peroxide-induced cell death
protein binding
protein disulfide oxidoreductase activity
peptide disulfide oxidoreductase activity
oxidoreductase activity, acting on a sulfur group of donors, disulfide as acceptor
poly(A) RNA binding
sulfate assimilation
negative regulation of transcription from RNA polymerase II promoter
response to reactive oxygen species
transcription, DNA-templated
protein folding
glycerol ether metabolic process
movement of cell or subcellular component
signal transduction
cell-cell signaling
cell proliferation
response to radiation
nucleobase-containing small molecule interconversion
protein repair
activation of protein kinase B activity
positive regulation of peptidyl-serine phosphorylation
regulation of protein import into nucleus, translocation
positive regulation of DNA binding
cell redox homeostasis
negative regulation of protein export from nucleus
positive regulation of protein kinase B signaling
oxidation-reduction process
negative regulation of hydrogen peroxide-induced cell death
protein binding
protein disulfide oxidoreductase activity
peptide disulfide oxidoreductase activity
oxidoreductase activity, acting on a sulfur group of donors, disulfide as acceptor
poly(A) RNA binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000012020.hg.1 | ENST00000441872.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012020.hg.1 | ENST00000441872.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018206.hg.1 | TC09001473.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018206.hg.1 | ENST00000374517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018206.hg.1 | NM_001244938.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018206.hg.1 | NM_003329.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014544.hg.1 | ENST00000443918.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018208.hg.1 | TC09001473.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018208.hg.1 | ENST00000374517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018208.hg.1 | NM_001244938.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018208.hg.1 | NM_003329.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018204.hg.1 | TC09001473.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018204.hg.1 | ENST00000374515.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018204.hg.1 | ENST00000374517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018204.hg.1 | NM_001244938.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018204.hg.1 | NM_003329.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018205.hg.1 | TC09001473.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018205.hg.1 | ENST00000374515.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018205.hg.1 | ENST00000374517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018205.hg.1 | NM_001244938.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09018205.hg.1 | NM_003329.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700006629.hg.1 | ENST00000575018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014577.hg.1 | ENST00000441043.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011177.hg.1 | ENST00000374515.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011177.hg.1 | ENST00000374515.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011177.hg.1 | ENST00000374515.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011177.hg.1 | ENST00000374515.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011177.hg.1 | ENST00000374515.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011177.hg.1 | ENST00000374515.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011177.hg.1 | ENST00000374517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011177.hg.1 | ENST00000374517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011177.hg.1 | ENST00000374517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011177.hg.1 | ENST00000374517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011177.hg.1 | ENST00000374517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011177.hg.1 | ENST00000374517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011177.hg.1 | ENST00000374517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011177.hg.1 | ENST00000374517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011177.hg.1 | ENST00000374517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011177.hg.1 | ENST00000374517.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011177.hg.1 | ENST00000487892.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011177.hg.1 | ENST00000487892.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011177.hg.1 | ENST00000487892.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011177.hg.1 | ENST00000487892.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011177.hg.1 | NM_001244938.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011177.hg.1 | NM_001244938.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011177.hg.1 | NM_001244938.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011177.hg.1 | NM_001244938.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011177.hg.1 | NM_001244938.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011177.hg.1 | NM_001244938.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011177.hg.1 | NM_001244938.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011177.hg.1 | NM_001244938.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011177.hg.1 | NM_001244938.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011177.hg.1 | NM_003329.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011177.hg.1 | NM_003329.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011177.hg.1 | NM_003329.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011177.hg.1 | NM_003329.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011177.hg.1 | NM_003329.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011177.hg.1 | NM_003329.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011177.hg.1 | NM_003329.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011177.hg.1 | NM_003329.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011177.hg.1 | NM_003329.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011177.hg.1 | NM_003329.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176025.hg.1 | ENST00000374517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900176025.hg.1 | NM_001244938.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176025.hg.1 | NM_003329.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176025.hg.1 | ENST00000374517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900176025.hg.1 | NM_001244938.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176025.hg.1 | NM_003329.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176025.hg.1 | TC0900011177.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900176028.hg.1 | ENST00000374517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900176028.hg.1 | NM_001244938.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176028.hg.1 | NM_003329.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176028.hg.1 | ENST00000374517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900176028.hg.1 | NM_001244938.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176028.hg.1 | NM_003329.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176028.hg.1 | TC0900011177.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100014544.hg.1 | ENST00000443918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900176027.hg.1 | ENST00000374517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900176027.hg.1 | NM_001244938.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176027.hg.1 | NM_003329.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176027.hg.1 | ENST00000374517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900176027.hg.1 | NM_001244938.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176027.hg.1 | NM_003329.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176027.hg.1 | TC0900011177.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200014577.hg.1 | ENST00000441043.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700006629.hg.1 | ENST00000575018.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900176021.hg.1 | ENST00000374515.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900176021.hg.1 | ENST00000374517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900176021.hg.1 | NM_001244938.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176021.hg.1 | NM_003329.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176021.hg.1 | ENST00000374515.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900176021.hg.1 | ENST00000374517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900176021.hg.1 | NM_001244938.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176021.hg.1 | NM_003329.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176021.hg.1 | TC0900011177.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900176024.hg.1 | ENST00000374515.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900176024.hg.1 | ENST00000374517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900176024.hg.1 | NM_001244938.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176024.hg.1 | NM_003329.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176024.hg.1 | ENST00000374515.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900176024.hg.1 | ENST00000374517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900176024.hg.1 | NM_001244938.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176024.hg.1 | NM_003329.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900176024.hg.1 | TC0900011177.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900011177.hg.1 | ENST00000374515.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011177.hg.1 | ENST00000374515.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011177.hg.1 | ENST00000374515.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011177.hg.1 | ENST00000374515.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011177.hg.1 | ENST00000374515.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011177.hg.1 | ENST00000374515.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011177.hg.1 | ENST00000374517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011177.hg.1 | ENST00000374517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011177.hg.1 | ENST00000374517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011177.hg.1 | ENST00000374517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011177.hg.1 | ENST00000374517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011177.hg.1 | ENST00000374517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011177.hg.1 | ENST00000374517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011177.hg.1 | ENST00000374517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011177.hg.1 | ENST00000374517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011177.hg.1 | ENST00000374517.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011177.hg.1 | ENST00000487892.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011177.hg.1 | ENST00000487892.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011177.hg.1 | ENST00000487892.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011177.hg.1 | ENST00000487892.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011177.hg.1 | NM_001244938.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011177.hg.1 | NM_001244938.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011177.hg.1 | NM_001244938.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011177.hg.1 | NM_001244938.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011177.hg.1 | NM_001244938.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011177.hg.1 | NM_001244938.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011177.hg.1 | NM_001244938.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011177.hg.1 | NM_001244938.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011177.hg.1 | NM_001244938.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011177.hg.1 | NM_003329.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011177.hg.1 | NM_003329.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011177.hg.1 | NM_003329.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011177.hg.1 | NM_003329.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011177.hg.1 | NM_003329.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011177.hg.1 | NM_003329.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011177.hg.1 | NM_003329.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011177.hg.1 | NM_003329.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011177.hg.1 | NM_003329.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011177.hg.1 | NM_003329.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012020.hg.1 | ENST00000441872.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012020.hg.1 | ENST00000441872.2 | Ensembl |

TXN
Genomic Map