Assay Details
| Gene Symbol: FOXD4L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012184.4 | NP_036316.1 | 1 | 1386 | 155 | |
| GenBank mRNA | AF452723.1 | - | 1 | 1234 | 155 | |
| AK122950.1 | - | 1 | 1386 | 155 | ||
| AK125398.1 | - | 1 | 1805 | 155 | ||
| AY344639.1 | - | 1 | 1298 | 155 | ||
| BC136428.1 | - | 1 | 1378 | 9020417 | 155 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000726.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 296789 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 88084 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 707538 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 88730 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 27336 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P119248 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15311164 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15311164 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 8161572 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8159811 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8155441 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8161584 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8155663 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8161585 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8155662 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8044639 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8161536 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8159812 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8161534 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8159813 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8161574 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8161586 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8159809 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8044637 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8159810 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8161573 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8044636 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8161535 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8044638 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8155664 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8161583 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8161533 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8155661 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8155440 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8044634 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8161571 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8159808 | - | - |
| Affymetrix Human Genome U219 Array | 11734409_PM_x_at | - | - |
| Affymetrix Human Genome U219 Array | 11734409_x_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009345.hg.1 | ENST00000382500.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009345.hg.1 | ENST00000382500.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009345.hg.1 | ENST00000382500.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009345.hg.1 | ENST00000382500.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009345.hg.1 | ENST00000382500.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009345.hg.1 | ENST00000382500.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009345.hg.1 | ENST00000382500.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009345.hg.1 | NM_207305.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009345.hg.1 | NM_207305.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009345.hg.1 | NM_207305.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009345.hg.1 | NM_207305.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009345.hg.1 | NM_207305.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009345.hg.1 | NM_207305.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009345.hg.1 | NM_207305.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009345.hg.1 | NM_207305.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02011321.hg.1 | TC02000726.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02011321.hg.1 | ENST00000306507.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02011321.hg.1 | NM_012184.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02011319.hg.1 | TC02000726.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02011319.hg.1 | ENST00000306507.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02011319.hg.1 | NM_012184.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008974.hg.1 | ENST00000306507.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008974.hg.1 | ENST00000306507.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008974.hg.1 | ENST00000306507.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008974.hg.1 | ENST00000306507.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008974.hg.1 | ENST00000306507.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008974.hg.1 | NM_012184.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008974.hg.1 | NM_012184.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008974.hg.1 | NM_012184.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008974.hg.1 | NM_012184.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02011320.hg.1 | TC02000726.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02011320.hg.1 | ENST00000306507.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02011320.hg.1 | NM_012184.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200162762.hg.1 | ENST00000306507.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200162762.hg.1 | NM_012184.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200162762.hg.1 | ENST00000306507.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200162762.hg.1 | NM_012184.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200162762.hg.1 | TC0200008974.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900009345.hg.1 | ENST00000382500.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009345.hg.1 | ENST00000382500.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009345.hg.1 | ENST00000382500.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009345.hg.1 | ENST00000382500.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009345.hg.1 | ENST00000382500.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009345.hg.1 | ENST00000382500.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009345.hg.1 | ENST00000382500.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009345.hg.1 | NM_207305.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009345.hg.1 | NM_207305.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009345.hg.1 | NM_207305.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009345.hg.1 | NM_207305.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009345.hg.1 | NM_207305.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009345.hg.1 | NM_207305.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009345.hg.1 | NM_207305.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009345.hg.1 | NM_207305.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200162758.hg.1 | ENST00000306507.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200162758.hg.1 | NM_012184.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200162758.hg.1 | ENST00000306507.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200162758.hg.1 | NM_012184.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200162758.hg.1 | TC0200008974.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200162760.hg.1 | ENST00000306507.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200162760.hg.1 | NM_012184.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200162760.hg.1 | ENST00000306507.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200162760.hg.1 | NM_012184.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200162760.hg.1 | TC0200008974.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200008974.hg.1 | ENST00000306507.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008974.hg.1 | ENST00000306507.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008974.hg.1 | ENST00000306507.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008974.hg.1 | ENST00000306507.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008974.hg.1 | ENST00000306507.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008974.hg.1 | NM_012184.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008974.hg.1 | NM_012184.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008974.hg.1 | NM_012184.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008974.hg.1 | NM_012184.4 | Refseq |
| Human Genome U133 Plus 2.0 Array | 217032_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 217032_at | - | - |
| Human Genome U133A 2.0 Array | 217032_at | - | - |
| Human Genome U133A 2.0 Array | 217032_PM_at | - | - |
| Human Genome U133 Set | 217032_at | - | - |
| Human Genome U133 Set | 217032_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A016230 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A037512 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A038740 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A037512 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A038740 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A016230 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 431174 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_2085441 | AK122950.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2085441 | AK125398.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2085441 | AY344639.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2085441 | NM_012184.4 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1781867 | BC136428.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1781867 | AK125398.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1781867 | AY344639.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1781867 | NM_012184.4 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1781867 | BC136428.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1781867 | AK125398.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1781867 | AY344639.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1781867 | NM_012184.4 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2085441 | AK122950.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2085441 | AK125398.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2085441 | AY344639.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2085441 | NM_012184.4 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1781867 | BC136428.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1781867 | AK125398.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1781867 | AY344639.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1781867 | NM_012184.4 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2085441 | AK122950.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2085441 | AK125398.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2085441 | AY344639.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2085441 | NM_012184.4 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2085441 | AK122950.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2085441 | AK125398.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2085441 | AY344639.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2085441 | NM_012184.4 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_45580705-S | BC136428.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_45580705-S | AK122950.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_45580705-S | AK125398.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_45580705-S | AY344639.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_45580705-S | NM_012184.4 | human_transcriptome |
| Illumina HumanRef-8 v2 Expression BeadChip | GI_3404-S | - | - |
| Illumina human-6 v1.0 expression beadchip | 0002370070 | - | - |
| Illumina humanRef-8 v1.0 expression beadchip | 0002370070 | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1781867 | BC136428.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1781867 | AK125398.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1781867 | AY344639.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1781867 | NM_012184.4 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1781867 | BC136428.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1781867 | AK125398.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1781867 | AY344639.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1781867 | NM_012184.4 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | ILMN_3404 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_3404 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2085441 | AK122950.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2085441 | AK125398.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2085441 | AY344639.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2085441 | NM_012184.4 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1781867 | BC136428.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1781867 | AK125398.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1781867 | AY344639.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1781867 | NM_012184.4 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2085441 | AK122950.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2085441 | AK125398.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2085441 | AY344639.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2085441 | NM_012184.4 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2085441 | AK122950.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2085441 | AK125398.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2085441 | AY344639.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2085441 | NM_012184.4 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1781867 | BC136428.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1781867 | AK125398.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1781867 | AY344639.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1781867 | NM_012184.4 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1781867 | BC136428.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1781867 | AK125398.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1781867 | AY344639.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1781867 | NM_012184.4 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2085441 | AK122950.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2085441 | AK125398.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2085441 | AY344639.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2085441 | NM_012184.4 | human_transcriptome |

FOXD4L1
Genomic Map