Assay Details
| Gene Symbol: PTTG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004219.3 | NP_004210.1 | 1 - 2 | 84 | 86 | |
| GenBank mRNA | AK312209.1 | - | 1 - 2 | 62 | 86 | |
| HY006026.1 | - | 1 - 2 | 85 | 86 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05000896.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK312209.1,NM_004219.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
DNA repair
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
mitotic nuclear division
spermatogenesis
negative regulation of endopeptidase activity
anaphase-promoting complex-dependent catabolic process
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
homologous chromosome segregation
chromosome organization
cell division
transcription factor activity, sequence-specific DNA binding
cysteine-type endopeptidase inhibitor activity
protein binding
SH3 domain binding
peptidase inhibitor activity
DNA repair
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
mitotic nuclear division
spermatogenesis
negative regulation of endopeptidase activity
anaphase-promoting complex-dependent catabolic process
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
homologous chromosome segregation
chromosome organization
cell division
transcription factor activity, sequence-specific DNA binding
cysteine-type endopeptidase inhibitor activity
protein binding
SH3 domain binding
peptidase inhibitor activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000352433.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000352433.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000352433.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000352433.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000352433.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000352433.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000352433.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000352433.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000352433.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000393964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000393964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000393964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000393964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000393964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000393964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000393964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000393964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000393964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000393964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000393964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000393964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000517480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000517480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000517480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000517480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000517480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000519287.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000519287.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000519287.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000519287.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000519287.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000519287.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000519287.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000519287.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000520452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000520452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000520452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000520452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000520452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000520452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000520452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000520452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000523659.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000523659.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000523659.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000523659.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000523659.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000523659.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000524244.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000524244.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000524244.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000524244.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_001282382.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_001282382.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_001282382.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_001282382.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_001282382.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_001282382.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_001282382.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_001282382.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_001282382.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_001282382.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_001282382.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_001282382.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_001282383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_001282383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_001282383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_001282383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_001282383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_001282383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_001282383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_001282383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_001282383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_004219.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_004219.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_004219.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_004219.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_004219.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_004219.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_004219.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_004219.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_004219.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05012880.hg.1 | TC05000896.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05012880.hg.1 | ENST00000352433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05012880.hg.1 | ENST00000393964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05012880.hg.1 | ENST00000520452.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05012880.hg.1 | NM_004219.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05012883.hg.1 | TC05000896.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05012883.hg.1 | ENST00000352433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05012883.hg.1 | ENST00000393964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05012883.hg.1 | ENST00000519287.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05012883.hg.1 | ENST00000520452.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05012883.hg.1 | NM_004219.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05012872.hg.1 | TC05000896.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05012872.hg.1 | ENST00000352433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05012872.hg.1 | NM_004219.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05012881.hg.1 | TC05000896.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05012881.hg.1 | ENST00000352433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05012881.hg.1 | ENST00000393964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05012881.hg.1 | ENST00000519287.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05012881.hg.1 | ENST00000520452.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05012881.hg.1 | NM_004219.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000352433.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000352433.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000352433.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000352433.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000352433.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000352433.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000352433.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000352433.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000352433.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000393964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000393964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000393964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000393964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000393964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000393964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000393964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000393964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000393964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000393964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000393964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000393964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000517480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000517480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000517480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000517480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000517480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000519287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000519287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000519287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000519287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000519287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000519287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000519287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000519287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000520452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000520452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000520452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000520452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000520452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000520452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000520452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000520452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000523659.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000523659.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000523659.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000523659.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000523659.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000523659.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000524244.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000524244.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000524244.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000524244.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_001282382.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_001282382.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_001282382.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_001282382.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_001282382.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_001282382.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_001282382.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_001282382.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_001282382.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_001282382.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_001282382.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_001282382.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_001282383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_001282383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_001282383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_001282383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_001282383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_001282383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_001282383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_001282383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_001282383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_004219.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_004219.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_004219.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_004219.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_004219.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_004219.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_004219.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_004219.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_004219.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500164830.hg.1 | ENST00000352433.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500164830.hg.1 | NM_004219.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500164830.hg.1 | ENST00000352433.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500164830.hg.1 | NM_004219.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500164830.hg.1 | TC0500009281.hg.1 | Clariom D TC |

PTTG1
Genomic Map