Assay Details
| Gene Symbol: TMPRSS11BNL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104048.1 | 2 - 3 | 287 | 102 | ||
| GenBank mRNA | AK123556.1 | - | 2 - 3 | 287 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400010900.hg.1 | ENST00000504453.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010900.hg.1 | ENST00000504453.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010900.hg.1 | ENST00000504453.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010900.hg.1 | ENST00000504453.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010900.hg.1 | ENST00000504453.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010900.hg.1 | ENST00000510782.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010900.hg.1 | ENST00000510782.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010900.hg.1 | ENST00000510782.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010900.hg.1 | ENST00000510782.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010900.hg.1 | ENST00000510782.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010900.hg.1 | ENST00000510782.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010900.hg.1 | ENST00000510782.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010900.hg.1 | ENST00000510782.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010900.hg.1 | ENST00000510782.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010900.hg.1 | ENST00000511720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010900.hg.1 | ENST00000511720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010900.hg.1 | ENST00000511720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010900.hg.1 | ENST00000511720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010900.hg.1 | ENST00000511720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010900.hg.1 | ENST00000511720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010900.hg.1 | ENST00000513714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010900.hg.1 | ENST00000513714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010900.hg.1 | ENST00000513714.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010900.hg.1 | ENST00000514295.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010900.hg.1 | ENST00000514295.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010900.hg.1 | ENST00000514295.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010900.hg.1 | ENST00000514295.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010900.hg.1 | ENST00000514295.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010900.hg.1 | ENST00000514295.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010900.hg.1 | ENST00000514295.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010900.hg.1 | ENST00000514295.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010900.hg.1 | ENST00000514295.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010900.hg.1 | ENST00000514295.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010900.hg.1 | ENST00000514295.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010900.hg.1 | ENST00000514295.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010900.hg.1 | ENST00000514295.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010900.hg.1 | ENST00000514295.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010900.hg.1 | ENST00000514420.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010900.hg.1 | ENST00000514420.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010900.hg.1 | ENST00000514420.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010900.hg.1 | NR_104048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010900.hg.1 | NR_104048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010900.hg.1 | NR_104048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010900.hg.1 | NR_104048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010900.hg.1 | NR_104048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010900.hg.1 | NR_104048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010900.hg.1 | NR_104048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010900.hg.1 | NR_104048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010900.hg.1 | NR_104048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010900.hg.1 | NR_104048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010900.hg.1 | NR_104048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010900.hg.1 | NR_104048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010900.hg.1 | NR_104048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010900.hg.1 | NR_104048.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010900.hg.1 | NR_104048.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400174132.hg.1 | ENST00000510782.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400174132.hg.1 | ENST00000511720.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400174132.hg.1 | ENST00000514295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400174132.hg.1 | NR_104048.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400174132.hg.1 | ENST00000510782.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400174132.hg.1 | ENST00000511720.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400174132.hg.1 | ENST00000514295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400174132.hg.1 | NR_104048.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400174132.hg.1 | TC0400010900.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400174131.hg.1 | ENST00000511720.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400174131.hg.1 | ENST00000514295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400174131.hg.1 | NR_104048.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400174131.hg.1 | ENST00000511720.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400174131.hg.1 | ENST00000514295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400174131.hg.1 | NR_104048.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400174131.hg.1 | TC0400010900.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400174136.hg.1 | ENST00000504453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400174136.hg.1 | ENST00000510782.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400174136.hg.1 | ENST00000514295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400174136.hg.1 | NR_104048.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400174136.hg.1 | ENST00000504453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400174136.hg.1 | ENST00000510782.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400174136.hg.1 | ENST00000514295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400174136.hg.1 | NR_104048.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400174136.hg.1 | TC0400010900.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400010900.hg.1 | ENST00000504453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010900.hg.1 | ENST00000504453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010900.hg.1 | ENST00000504453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010900.hg.1 | ENST00000504453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010900.hg.1 | ENST00000504453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010900.hg.1 | ENST00000510782.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010900.hg.1 | ENST00000510782.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010900.hg.1 | ENST00000510782.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010900.hg.1 | ENST00000510782.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010900.hg.1 | ENST00000510782.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010900.hg.1 | ENST00000510782.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010900.hg.1 | ENST00000510782.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010900.hg.1 | ENST00000510782.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010900.hg.1 | ENST00000510782.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010900.hg.1 | ENST00000511720.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010900.hg.1 | ENST00000511720.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010900.hg.1 | ENST00000511720.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010900.hg.1 | ENST00000511720.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010900.hg.1 | ENST00000511720.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010900.hg.1 | ENST00000511720.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010900.hg.1 | ENST00000513714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010900.hg.1 | ENST00000513714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010900.hg.1 | ENST00000513714.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010900.hg.1 | ENST00000514295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010900.hg.1 | ENST00000514295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010900.hg.1 | ENST00000514295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010900.hg.1 | ENST00000514295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010900.hg.1 | ENST00000514295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010900.hg.1 | ENST00000514295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010900.hg.1 | ENST00000514295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010900.hg.1 | ENST00000514295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010900.hg.1 | ENST00000514295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010900.hg.1 | ENST00000514295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010900.hg.1 | ENST00000514295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010900.hg.1 | ENST00000514295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010900.hg.1 | ENST00000514295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010900.hg.1 | ENST00000514295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010900.hg.1 | ENST00000514420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010900.hg.1 | ENST00000514420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010900.hg.1 | ENST00000514420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400010900.hg.1 | NR_104048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010900.hg.1 | NR_104048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010900.hg.1 | NR_104048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010900.hg.1 | NR_104048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010900.hg.1 | NR_104048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010900.hg.1 | NR_104048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010900.hg.1 | NR_104048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010900.hg.1 | NR_104048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010900.hg.1 | NR_104048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010900.hg.1 | NR_104048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010900.hg.1 | NR_104048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010900.hg.1 | NR_104048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010900.hg.1 | NR_104048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010900.hg.1 | NR_104048.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400010900.hg.1 | NR_104048.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400174137.hg.1 | ENST00000510782.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400174137.hg.1 | ENST00000514295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400174137.hg.1 | NR_104048.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400174137.hg.1 | ENST00000510782.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400174137.hg.1 | ENST00000514295.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400174137.hg.1 | NR_104048.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400174137.hg.1 | TC0400010900.hg.1 | Clariom D TC |

TMPRSS11BNL
Genomic Map