Assay Details
| Gene Symbol: VKORC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001311311.1 | NP_001298240.1 | 1 - 2 | 448 | 51 | |
| NM_024006.5 | NP_076869.1 | 1 - 2 | 448 | 51 | ||
| GenBank mRNA | AF176924.1 | - | 1 - 2 | 390 | 51 | |
| AK312005.1 | - | 1 - 2 | 361 | 51 | ||
| AY423044.1 | - | 1 - 2 | 178 | 51 | ||
| AY466113.1 | - | 1 - 2 | 390 | 51 | ||
| AY521634.1 | - | 1 - 2 | 178 | 51 | ||
| BC000828.1 | - | 1 - 2 | 178 | 51 | ||
| BC002911.2 | - | 1 - 2 | 216 | 3944530 | 51 | |
| BI822140.1 | - | 1 - 2 | 404 | 51 | ||
| CN484751.1 | - | 1 - 2 | 175 | 51 | ||
| KU178762.1 | - | 1 - 2 | 178 | 51 | ||
More Information
Set Membership: |
|

VKORC1
Genomic Map