| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011168.hg.1 |
ENST00000542219.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011168.hg.1 |
ENST00000542219.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011168.hg.1 |
ENST00000542219.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011168.hg.1 |
ENST00000542219.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011168.hg.1 |
ENST00000542219.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011168.hg.1 |
ENST00000542219.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011168.hg.1 |
ENST00000542219.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011168.hg.1 |
ENST00000542219.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011168.hg.1 |
ENST00000542219.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011168.hg.1 |
ENST00000542219.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011168.hg.1 |
NR_110063.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011168.hg.1 |
NR_110063.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011168.hg.1 |
NR_110063.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011168.hg.1 |
NR_110063.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011168.hg.1 |
NR_110063.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011168.hg.1 |
NR_110063.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011168.hg.1 |
NR_110063.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011168.hg.1 |
NR_110063.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011168.hg.1 |
NR_110063.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011168.hg.1 |
NR_110063.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200186658.hg.1 |
ENST00000542219.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200186658.hg.1 |
NR_110063.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200186658.hg.1 |
ENST00000542219.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200186658.hg.1 |
NR_110063.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200186658.hg.1 |
TC1200011168.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1200011168.hg.1 |
ENST00000542219.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011168.hg.1 |
ENST00000542219.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011168.hg.1 |
ENST00000542219.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011168.hg.1 |
ENST00000542219.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011168.hg.1 |
ENST00000542219.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011168.hg.1 |
ENST00000542219.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011168.hg.1 |
ENST00000542219.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011168.hg.1 |
ENST00000542219.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011168.hg.1 |
ENST00000542219.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011168.hg.1 |
ENST00000542219.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011168.hg.1 |
NR_110063.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011168.hg.1 |
NR_110063.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011168.hg.1 |
NR_110063.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011168.hg.1 |
NR_110063.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011168.hg.1 |
NR_110063.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011168.hg.1 |
NR_110063.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011168.hg.1 |
NR_110063.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011168.hg.1 |
NR_110063.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011168.hg.1 |
NR_110063.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011168.hg.1 |
NR_110063.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200186657.hg.1 |
ENST00000542219.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200186657.hg.1 |
NR_110063.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200186657.hg.1 |
ENST00000542219.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200186657.hg.1 |
NR_110063.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200186657.hg.1 |
TC1200011168.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1200186656.hg.1 |
ENST00000542219.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200186656.hg.1 |
NR_110063.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200186656.hg.1 |
ENST00000542219.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200186656.hg.1 |
NR_110063.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200186656.hg.1 |
TC1200011168.hg.1 |
Clariom D TC |