Assay Details
| Gene Symbol: LOC101928288 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AL442080.1 | - | 1 - 2 | 193 | 89 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 6082 | MIR6082 | hsa-mir-6082 | -103.2kb | Anti-sense | Chr. 4 - 171186184 - 171186292 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC441052 | LOC441052 | 513.9kb | Sense | Chr. 4 - 171812138 - 171813607 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC101928288
Genomic Map