Assay Details
| Gene Symbol: LOC101927045 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110032.1 | 1 - 2 | 186 | 99 | ||
| GenBank mRNA | AK056368.1 | - | 1 - 2 | 167 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

LOC101927045
Genomic Map