Assay Details
| Gene Symbol: LINC00704 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_024475.1 | 2 - 3 | 248 | 76 | ||
| GenBank mRNA | BC048320.1 | - | 2 - 3 | 248 | 76 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000982.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10012334.hg.1 | ENST00000430998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10012334.hg.1 | NR_024475.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10012334.hg.1 | TC10000982.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10012335.hg.1 | ENST00000430998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10012335.hg.1 | NR_024475.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10012335.hg.1 | TC10000982.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009642.hg.1 | ENST00000430998.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009642.hg.1 | ENST00000430998.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009642.hg.1 | ENST00000430998.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009642.hg.1 | ENST00000430998.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009642.hg.1 | ENST00000430998.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009642.hg.1 | ENST00000430998.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009642.hg.1 | ENST00000430998.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009642.hg.1 | ENST00000430998.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009642.hg.1 | ENST00000430998.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009642.hg.1 | ENST00000430998.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009642.hg.1 | ENST00000430998.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009642.hg.1 | ENST00000430998.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009642.hg.1 | ENST00000430998.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009642.hg.1 | ENST00000430998.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009642.hg.1 | ENST00000443994.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009642.hg.1 | ENST00000443994.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009642.hg.1 | ENST00000443994.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009642.hg.1 | ENST00000443994.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009642.hg.1 | ENST00000449712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009642.hg.1 | ENST00000449712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009642.hg.1 | ENST00000449712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009642.hg.1 | ENST00000449712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009642.hg.1 | ENST00000449712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009642.hg.1 | ENST00000449712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009642.hg.1 | NR_024475.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009642.hg.1 | NR_024475.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009642.hg.1 | NR_024475.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009642.hg.1 | NR_024475.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009642.hg.1 | NR_024475.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000009642.hg.1 | NR_024475.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000165857.hg.1 | ENST00000430998.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000165857.hg.1 | ENST00000430998.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000165857.hg.1 | ENST00000449712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000165857.hg.1 | NR_024475.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000165857.hg.1 | ENST00000430998.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000165857.hg.1 | ENST00000430998.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000165857.hg.1 | ENST00000449712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000165857.hg.1 | NR_024475.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000165857.hg.1 | TC1000009642.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000165859.hg.1 | ENST00000430998.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000165859.hg.1 | ENST00000430998.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000165859.hg.1 | ENST00000449712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000165859.hg.1 | NR_024475.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000165859.hg.1 | ENST00000430998.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000165859.hg.1 | ENST00000430998.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000165859.hg.1 | ENST00000449712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000165859.hg.1 | NR_024475.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000165859.hg.1 | TC1000009642.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000009642.hg.1 | ENST00000430998.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009642.hg.1 | ENST00000430998.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009642.hg.1 | ENST00000430998.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009642.hg.1 | ENST00000430998.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009642.hg.1 | ENST00000430998.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009642.hg.1 | ENST00000430998.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009642.hg.1 | ENST00000430998.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009642.hg.1 | ENST00000430998.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009642.hg.1 | ENST00000430998.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009642.hg.1 | ENST00000430998.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009642.hg.1 | ENST00000430998.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009642.hg.1 | ENST00000430998.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009642.hg.1 | ENST00000430998.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009642.hg.1 | ENST00000430998.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009642.hg.1 | ENST00000443994.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009642.hg.1 | ENST00000443994.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009642.hg.1 | ENST00000443994.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009642.hg.1 | ENST00000443994.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009642.hg.1 | ENST00000449712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009642.hg.1 | ENST00000449712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009642.hg.1 | ENST00000449712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009642.hg.1 | ENST00000449712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009642.hg.1 | ENST00000449712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009642.hg.1 | ENST00000449712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000009642.hg.1 | NR_024475.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009642.hg.1 | NR_024475.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009642.hg.1 | NR_024475.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009642.hg.1 | NR_024475.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009642.hg.1 | NR_024475.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000009642.hg.1 | NR_024475.1 | Refseq |

LINC00704
Genomic Map