Assay Details
| Gene Symbol: MMD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270375.1 | NP_001257304.1 | 5 - 6 | 664 | 62 | |
| NM_198403.3 | NP_940685.3 | 5 - 6 | 664 | 62 | ||
| GenBank mRNA | AY424288.1 | - | 5 - 6 | 573 | 62 | |
| BC067905.1 | - | 5 - 6 | 664 | 5312325 | 62 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07001113.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR07017105.hg.1 | TC07001113.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07017105.hg.1 | ENST00000401401.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07017105.hg.1 | ENST00000404774.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07017105.hg.1 | ENST00000406755.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07017105.hg.1 | NM_001100600.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07017105.hg.1 | NM_198403.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | ENST00000401401.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | ENST00000401401.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | ENST00000401401.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | ENST00000401401.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | ENST00000401401.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | ENST00000401401.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | ENST00000401401.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | ENST00000401401.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | ENST00000401401.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | ENST00000401401.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | ENST00000401401.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | ENST00000404774.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | ENST00000404774.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | ENST00000404774.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | ENST00000404774.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | ENST00000404774.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | ENST00000404774.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | ENST00000404774.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | ENST00000404774.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | ENST00000404774.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | ENST00000404774.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | ENST00000404774.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | ENST00000404774.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | ENST00000406755.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | ENST00000406755.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | ENST00000406755.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | ENST00000406755.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | ENST00000406755.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | ENST00000406755.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | ENST00000406755.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | ENST00000406755.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | ENST00000406755.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | ENST00000406755.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | ENST00000406755.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | ENST00000406755.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | ENST00000612910.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | ENST00000612910.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | ENST00000612910.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | ENST00000612910.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | ENST00000612910.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | ENST00000612910.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | ENST00000612910.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | ENST00000612910.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | ENST00000612910.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | NM_001100600.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | NM_001100600.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | NM_001100600.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | NM_001100600.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | NM_001100600.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | NM_001100600.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | NM_001100600.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | NM_001100600.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | NM_001100600.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | NM_001100600.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | NM_001100600.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | NM_001100600.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | NM_001270375.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | NM_001270375.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | NM_001270375.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | NM_001270375.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | NM_001270375.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | NM_001270375.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | NM_001270375.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | NM_001270375.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | NM_001270375.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | NM_001270375.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | NM_001270375.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | NM_001270375.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | NM_198403.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | NM_198403.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | NM_198403.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | NM_198403.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | NM_198403.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | NM_198403.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | NM_198403.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | NM_198403.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | NM_198403.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | NM_198403.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | NM_198403.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | NR_072989.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | NR_072989.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | NR_072989.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | NR_072989.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | NR_072989.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | NR_072989.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | NR_072989.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | NR_072989.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | NR_072989.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | NR_072989.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | NR_072989.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010141.hg.1 | NR_072989.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | ENST00000401401.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | ENST00000401401.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | ENST00000401401.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | ENST00000401401.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | ENST00000401401.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | ENST00000401401.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | ENST00000401401.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | ENST00000401401.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | ENST00000401401.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | ENST00000401401.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | ENST00000401401.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | ENST00000404774.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | ENST00000404774.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | ENST00000404774.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | ENST00000404774.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | ENST00000404774.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | ENST00000404774.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | ENST00000404774.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | ENST00000404774.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | ENST00000404774.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | ENST00000404774.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | ENST00000404774.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | ENST00000404774.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | ENST00000406755.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | ENST00000406755.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | ENST00000406755.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | ENST00000406755.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | ENST00000406755.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | ENST00000406755.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | ENST00000406755.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | ENST00000406755.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | ENST00000406755.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | ENST00000406755.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | ENST00000406755.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | ENST00000406755.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | ENST00000612910.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | ENST00000612910.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | ENST00000612910.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | ENST00000612910.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | ENST00000612910.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | ENST00000612910.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | ENST00000612910.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | ENST00000612910.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | ENST00000612910.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | NM_001100600.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | NM_001100600.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | NM_001100600.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | NM_001100600.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | NM_001100600.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | NM_001100600.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | NM_001100600.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | NM_001100600.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | NM_001100600.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | NM_001100600.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | NM_001100600.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | NM_001100600.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | NM_001270375.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | NM_001270375.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | NM_001270375.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | NM_001270375.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | NM_001270375.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | NM_001270375.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | NM_001270375.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | NM_001270375.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | NM_001270375.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | NM_001270375.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | NM_001270375.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | NM_001270375.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | NM_198403.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | NM_198403.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | NM_198403.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | NM_198403.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | NM_198403.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | NM_198403.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | NM_198403.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | NM_198403.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | NM_198403.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | NM_198403.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | NM_198403.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | NR_072989.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | NR_072989.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | NR_072989.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | NR_072989.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | NR_072989.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | NR_072989.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | NR_072989.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | NR_072989.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | NR_072989.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | NR_072989.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | NR_072989.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010141.hg.1 | NR_072989.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700172048.hg.1 | ENST00000406755.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700172048.hg.1 | ENST00000612910.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700172048.hg.1 | NM_001270375.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700172048.hg.1 | ENST00000406755.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700172048.hg.1 | ENST00000612910.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700172048.hg.1 | NM_001270375.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700172048.hg.1 | TC0700010141.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700172049.hg.1 | ENST00000401401.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700172049.hg.1 | ENST00000404774.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700172049.hg.1 | ENST00000406755.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700172049.hg.1 | ENST00000612910.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700172049.hg.1 | NM_001100600.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700172049.hg.1 | NM_001270375.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700172049.hg.1 | NM_198403.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700172049.hg.1 | NR_072989.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700172049.hg.1 | ENST00000401401.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700172049.hg.1 | ENST00000404774.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700172049.hg.1 | ENST00000406755.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700172049.hg.1 | ENST00000612910.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700172049.hg.1 | NM_001100600.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700172049.hg.1 | NM_001270375.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700172049.hg.1 | NM_198403.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700172049.hg.1 | NR_072989.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700172049.hg.1 | TC0700010141.hg.1 | Clariom D TC |

MMD2
Genomic Map