Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005237.hg.1 |
ENST00000328264.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005237.hg.1 |
ENST00000398948.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005237.hg.1 |
NM_005867.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005237.hg.1 |
TC21000445.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005236.hg.1 |
ENST00000328264.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005236.hg.1 |
ENST00000398948.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005236.hg.1 |
NM_005867.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21005236.hg.1 |
TC21000445.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008128.hg.1 |
ENST00000328264.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008128.hg.1 |
ENST00000328264.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008128.hg.1 |
ENST00000328264.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008128.hg.1 |
ENST00000328264.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008128.hg.1 |
ENST00000328264.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008128.hg.1 |
ENST00000328264.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008128.hg.1 |
ENST00000328264.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008128.hg.1 |
ENST00000398948.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008128.hg.1 |
ENST00000398948.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008128.hg.1 |
ENST00000398948.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008128.hg.1 |
ENST00000398948.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008128.hg.1 |
ENST00000398948.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008128.hg.1 |
ENST00000482032.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008128.hg.1 |
ENST00000482032.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008128.hg.1 |
ENST00000482032.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008128.hg.1 |
ENST00000482032.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008128.hg.1 |
ENST00000482032.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008128.hg.1 |
ENST00000482032.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008128.hg.1 |
ENST00000482032.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008128.hg.1 |
NM_005867.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008128.hg.1 |
NM_005867.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008128.hg.1 |
NM_005867.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008128.hg.1 |
NM_005867.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008128.hg.1 |
NM_005867.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008128.hg.1 |
NM_005867.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100008128.hg.1 |
NM_005867.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100154790.hg.1 |
ENST00000328264.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154790.hg.1 |
ENST00000398948.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154790.hg.1 |
ENST00000482032.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154790.hg.1 |
NM_005867.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100154790.hg.1 |
TC2100008128.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100154790.hg.1 |
ENST00000328264.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154790.hg.1 |
ENST00000398948.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154790.hg.1 |
ENST00000482032.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154790.hg.1 |
NM_005867.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008128.hg.1 |
ENST00000328264.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008128.hg.1 |
ENST00000328264.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008128.hg.1 |
ENST00000328264.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008128.hg.1 |
ENST00000328264.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008128.hg.1 |
ENST00000328264.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008128.hg.1 |
ENST00000328264.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008128.hg.1 |
ENST00000328264.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008128.hg.1 |
ENST00000398948.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008128.hg.1 |
ENST00000398948.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008128.hg.1 |
ENST00000398948.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008128.hg.1 |
ENST00000398948.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008128.hg.1 |
ENST00000398948.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008128.hg.1 |
ENST00000482032.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008128.hg.1 |
ENST00000482032.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008128.hg.1 |
ENST00000482032.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008128.hg.1 |
ENST00000482032.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008128.hg.1 |
ENST00000482032.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008128.hg.1 |
ENST00000482032.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008128.hg.1 |
ENST00000482032.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100008128.hg.1 |
NM_005867.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008128.hg.1 |
NM_005867.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008128.hg.1 |
NM_005867.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008128.hg.1 |
NM_005867.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008128.hg.1 |
NM_005867.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008128.hg.1 |
NM_005867.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100008128.hg.1 |
NM_005867.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100154791.hg.1 |
ENST00000328264.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154791.hg.1 |
ENST00000398948.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154791.hg.1 |
ENST00000482032.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154791.hg.1 |
NM_005867.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100154791.hg.1 |
TC2100008128.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100154791.hg.1 |
ENST00000328264.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154791.hg.1 |
ENST00000398948.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154791.hg.1 |
ENST00000482032.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154791.hg.1 |
NM_005867.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100154792.hg.1 |
ENST00000328264.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154792.hg.1 |
ENST00000398948.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154792.hg.1 |
ENST00000482032.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154792.hg.1 |
NM_005867.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100154792.hg.1 |
TC2100008128.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100154792.hg.1 |
ENST00000328264.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154792.hg.1 |
ENST00000398948.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154792.hg.1 |
ENST00000482032.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100154792.hg.1 |
NM_005867.3 |
Refseq |