Assay Details
| Gene Symbol: LOC101928460 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110758.1 | 2 - 3 | 573 | 99 | ||
| GenBank mRNA | BC040893.1 | - | 2 - 3 | 573 | 99 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
protein tyrosine phosphatase, receptor type U |
PTPRU |
FMI, PCP-2, PTP, PTP-J, PTP-PI, PTP-RO, PTPPSI, PTPRO, PTPU2, R-PTP-PSI, R-PTP-U, hPTP-J |
-2.8kb | Anti-sense | Chr. 1 - 29236516 - 29326813 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC101929406 | LOC101929406 | 680.6kb | Sense | Chr. 1 - 30013952 - 30037612 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC101928460
Genomic Map