| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008205.hg.1 |
ENST00000623604.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800008205.hg.1 |
ENST00000623604.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21000484.hg.1 |
ENST00000324988.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21000484.hg.1 |
NR_027246.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21000484.hg.1 |
TC21000090.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006833.hg.1 |
ENST00000324988.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006833.hg.1 |
ENST00000324988.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006833.hg.1 |
ENST00000324988.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006833.hg.1 |
NR_027246.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006833.hg.1 |
NR_027246.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006833.hg.1 |
NR_027246.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21000485.hg.1 |
ENST00000324988.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21000485.hg.1 |
NR_027246.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21000485.hg.1 |
TC21000090.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006545.hg.1 |
ENST00000624445.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006545.hg.1 |
ENST00000624445.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100145990.hg.1 |
ENST00000324988.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100145990.hg.1 |
NR_027246.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100145990.hg.1 |
TC2100006833.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2100145990.hg.1 |
ENST00000324988.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100145990.hg.1 |
NR_027246.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100145991.hg.1 |
ENST00000324988.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100145991.hg.1 |
NR_027246.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100145991.hg.1 |
TC2100006833.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2100145991.hg.1 |
ENST00000324988.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100145991.hg.1 |
NR_027246.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100006545.hg.1 |
ENST00000624445.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006545.hg.1 |
ENST00000624445.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008205.hg.1 |
ENST00000623604.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800008205.hg.1 |
ENST00000623604.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006833.hg.1 |
ENST00000324988.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006833.hg.1 |
ENST00000324988.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006833.hg.1 |
ENST00000324988.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006833.hg.1 |
NR_027246.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100006833.hg.1 |
NR_027246.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100006833.hg.1 |
NR_027246.1 |
Refseq |