Assay Details
| Gene Symbol: LOC101928271 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC042077.1 | - | 1 - 2 | 191 | 105 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | lipid droplet associated hydrolase | LDAH |
C2orf43, hLDAH |
-36.6kb | Anti-sense | Chr. 2 - 20682489 - 20823139 on NCBI Build GRCh38 |
| Downstream | apolipoprotein B | APOB |
FLDB, LDLCQ4, apoB-100, apoB-48 |
137.6kb | Anti-sense | Chr. 2 - 21001429 - 21044073 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC101928271
Genomic Map