Assay Details
| Gene Symbol: HLA-DQA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK130598.1 | - | 4 - 5 | 830 | 152 | |
| AK130838.1 | - | 4 - 5 | 832 | 152 | ||
| BC008585.1 | - | 4 - 5 | 813 | 4339446 | 152 | |
| M20506.1 | - | 3 - 4 | 678 | 152 | ||
| X00370.1 | - | 4 - 5 | 789 | 152 | ||
More Information
Set Membership: |
|

HLA-DQA1
Genomic Map