Assay Details
| Gene Symbol: NAPSB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002798.1 | 3 - 4 | 346 | 131 | ||
| GenBank mRNA | AF090387.1 | - | 3 - 4 | 353 | 131 | |
| AF098485.1 | - | 3 - 4 | 353 | 131 | ||
| AF200344.1 | - | 3 - 4 | 354 | 131 | ||
| AK300884.1 | - | 3 - 4 | 357 | 131 | ||
| AK310134.1 | - | 3 - 4 | 364 | 131 | ||
| BC085008.1 | - | 3 - 4 | 299 | 131 | ||
| BC111705.1 | - | 3 - 4 | 343 | 131 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001745.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900011195.hg.1 | ENST00000525179.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011195.hg.1 | ENST00000525179.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011195.hg.1 | ENST00000525179.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011195.hg.1 | ENST00000525179.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011195.hg.1 | ENST00000525179.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011195.hg.1 | ENST00000527780.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011195.hg.1 | ENST00000527780.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011195.hg.1 | ENST00000527780.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011195.hg.1 | ENST00000527780.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011195.hg.1 | ENST00000527780.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011195.hg.1 | ENST00000527780.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011195.hg.1 | ENST00000527780.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011195.hg.1 | ENST00000527780.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011195.hg.1 | ENST00000527780.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011195.hg.1 | ENST00000527780.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011195.hg.1 | ENST00000531692.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011195.hg.1 | ENST00000531692.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011195.hg.1 | ENST00000531692.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011195.hg.1 | ENST00000531692.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011195.hg.1 | ENST00000534096.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011195.hg.1 | ENST00000534096.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011195.hg.1 | ENST00000534096.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011195.hg.1 | ENST00000534096.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011195.hg.1 | ENST00000534096.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011195.hg.1 | ENST00000534096.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011195.hg.1 | ENST00000534096.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011195.hg.1 | ENST00000534096.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011195.hg.1 | ENST00000534789.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011195.hg.1 | ENST00000534789.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011195.hg.1 | ENST00000534789.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011195.hg.1 | ENST00000534789.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011195.hg.1 | ENST00000534789.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011195.hg.1 | ENST00000562112.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011195.hg.1 | ENST00000562112.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011195.hg.1 | ENST00000562112.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011195.hg.1 | ENST00000562112.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011195.hg.1 | ENST00000562112.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011195.hg.1 | ENST00000562112.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011195.hg.1 | ENST00000562112.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011195.hg.1 | ENST00000562112.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011195.hg.1 | ENST00000562112.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011195.hg.1 | NR_002798.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011195.hg.1 | NR_002798.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011195.hg.1 | NR_002798.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011195.hg.1 | NR_002798.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011195.hg.1 | NR_002798.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011195.hg.1 | NR_002798.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011195.hg.1 | NR_002798.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011195.hg.1 | NR_002798.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011195.hg.1 | NR_002798.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011195.hg.1 | NR_002798.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023758.hg.1 | ENST00000527780.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023758.hg.1 | ENST00000534096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023758.hg.1 | NR_002798.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023758.hg.1 | TC19001745.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023759.hg.1 | ENST00000527780.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023759.hg.1 | ENST00000534096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023759.hg.1 | NR_002798.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023759.hg.1 | TC19001745.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023761.hg.1 | ENST00000525179.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023761.hg.1 | ENST00000527780.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023761.hg.1 | ENST00000534096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023761.hg.1 | NR_002798.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19023761.hg.1 | TC19001745.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1900194007.hg.1 | ENST00000527780.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194007.hg.1 | ENST00000534096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194007.hg.1 | ENST00000562112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194007.hg.1 | NR_002798.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900194007.hg.1 | TC1900011195.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900194007.hg.1 | ENST00000527780.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194007.hg.1 | ENST00000534096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194007.hg.1 | ENST00000562112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194007.hg.1 | NR_002798.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900194010.hg.1 | ENST00000525179.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194010.hg.1 | ENST00000527780.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194010.hg.1 | ENST00000534096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194010.hg.1 | ENST00000562112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194010.hg.1 | NR_002798.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900194010.hg.1 | TC1900011195.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900194010.hg.1 | ENST00000525179.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194010.hg.1 | ENST00000527780.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194010.hg.1 | ENST00000534096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194010.hg.1 | ENST00000562112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194010.hg.1 | NR_002798.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011195.hg.1 | ENST00000525179.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011195.hg.1 | ENST00000525179.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011195.hg.1 | ENST00000525179.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011195.hg.1 | ENST00000525179.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011195.hg.1 | ENST00000525179.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011195.hg.1 | ENST00000527780.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011195.hg.1 | ENST00000527780.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011195.hg.1 | ENST00000527780.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011195.hg.1 | ENST00000527780.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011195.hg.1 | ENST00000527780.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011195.hg.1 | ENST00000527780.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011195.hg.1 | ENST00000527780.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011195.hg.1 | ENST00000527780.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011195.hg.1 | ENST00000527780.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011195.hg.1 | ENST00000527780.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011195.hg.1 | ENST00000531692.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011195.hg.1 | ENST00000531692.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011195.hg.1 | ENST00000531692.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011195.hg.1 | ENST00000531692.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011195.hg.1 | ENST00000534096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011195.hg.1 | ENST00000534096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011195.hg.1 | ENST00000534096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011195.hg.1 | ENST00000534096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011195.hg.1 | ENST00000534096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011195.hg.1 | ENST00000534096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011195.hg.1 | ENST00000534096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011195.hg.1 | ENST00000534096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011195.hg.1 | ENST00000534789.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011195.hg.1 | ENST00000534789.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011195.hg.1 | ENST00000534789.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011195.hg.1 | ENST00000534789.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011195.hg.1 | ENST00000534789.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011195.hg.1 | ENST00000562112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011195.hg.1 | ENST00000562112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011195.hg.1 | ENST00000562112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011195.hg.1 | ENST00000562112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011195.hg.1 | ENST00000562112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011195.hg.1 | ENST00000562112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011195.hg.1 | ENST00000562112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011195.hg.1 | ENST00000562112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011195.hg.1 | ENST00000562112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011195.hg.1 | NR_002798.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011195.hg.1 | NR_002798.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011195.hg.1 | NR_002798.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011195.hg.1 | NR_002798.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011195.hg.1 | NR_002798.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011195.hg.1 | NR_002798.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011195.hg.1 | NR_002798.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011195.hg.1 | NR_002798.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011195.hg.1 | NR_002798.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011195.hg.1 | NR_002798.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900194008.hg.1 | ENST00000527780.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194008.hg.1 | ENST00000534096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194008.hg.1 | ENST00000562112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194008.hg.1 | NR_002798.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900194008.hg.1 | TC1900011195.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900194008.hg.1 | ENST00000527780.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194008.hg.1 | ENST00000534096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194008.hg.1 | ENST00000562112.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900194008.hg.1 | NR_002798.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 141392 | - | - |

NAPSB
Genomic Map