Assay Details
Gene Symbol: ZNF528-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_125345.1 | 1 - 2 | 299 | 153 | ||
GenBank mRNA | BI755704.1 | - | 1 - 2 | 308 | 153 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000594119.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000594119.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000594119.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000594119.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000594119.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000596746.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000596746.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000596746.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000596746.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000596746.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000596746.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000596746.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000596746.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000596746.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000596746.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000596746.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000598892.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000598892.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000598892.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000598892.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | NR_125345.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | NR_125345.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | NR_125345.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | NR_125345.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | NR_125345.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | NR_125345.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | NR_125345.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | NR_125345.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | NR_125345.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | NR_125345.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | NR_125345.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011295.hg.1 | NR_125345.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900195273.hg.1 | ENST00000596746.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195273.hg.1 | ENST00000598892.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195273.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195273.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195273.hg.1 | NR_125345.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900195273.hg.1 | TC1900011295.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900195273.hg.1 | ENST00000596746.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195273.hg.1 | ENST00000598892.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195273.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195273.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195273.hg.1 | NR_125345.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900195259.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195259.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195259.hg.1 | NR_125345.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900195259.hg.1 | TC1900011295.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900195259.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195259.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195259.hg.1 | NR_125345.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900195263.hg.1 | ENST00000596746.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195263.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195263.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195263.hg.1 | NR_125345.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900195263.hg.1 | TC1900011295.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900195263.hg.1 | ENST00000596746.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195263.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195263.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195263.hg.1 | NR_125345.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900195266.hg.1 | ENST00000596746.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195266.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195266.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195266.hg.1 | NR_125345.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900195266.hg.1 | TC1900011295.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900195266.hg.1 | ENST00000596746.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195266.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195266.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195266.hg.1 | NR_125345.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900195269.hg.1 | ENST00000596746.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195269.hg.1 | ENST00000598892.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195269.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195269.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195269.hg.1 | NR_125345.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900195269.hg.1 | TC1900011295.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900195269.hg.1 | ENST00000596746.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195269.hg.1 | ENST00000598892.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195269.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195269.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195269.hg.1 | NR_125345.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000594119.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000594119.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000594119.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000594119.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000594119.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000596746.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000596746.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000596746.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000596746.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000596746.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000596746.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000596746.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000596746.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000596746.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000596746.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000596746.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000598892.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000598892.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000598892.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000598892.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | NR_125345.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | NR_125345.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | NR_125345.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | NR_125345.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | NR_125345.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | NR_125345.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | NR_125345.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | NR_125345.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | NR_125345.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | NR_125345.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | NR_125345.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011295.hg.1 | NR_125345.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900195268.hg.1 | ENST00000596746.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195268.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195268.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195268.hg.1 | NR_125345.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900195268.hg.1 | TC1900011295.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900195268.hg.1 | ENST00000596746.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195268.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195268.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195268.hg.1 | NR_125345.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900195274.hg.1 | ENST00000596746.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195274.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195274.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195274.hg.1 | NR_125345.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900195274.hg.1 | TC1900011295.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900195274.hg.1 | ENST00000596746.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195274.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195274.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195274.hg.1 | NR_125345.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900195251.hg.1 | ENST00000594119.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195251.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195251.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195251.hg.1 | NR_125345.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900195251.hg.1 | TC1900011295.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900195251.hg.1 | ENST00000594119.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195251.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195251.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195251.hg.1 | NR_125345.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900195262.hg.1 | ENST00000596746.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195262.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195262.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195262.hg.1 | NR_125345.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900195262.hg.1 | TC1900011295.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900195262.hg.1 | ENST00000596746.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195262.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195262.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195262.hg.1 | NR_125345.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900195250.hg.1 | ENST00000594119.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195250.hg.1 | NR_125345.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900195250.hg.1 | TC1900011295.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900195250.hg.1 | ENST00000594119.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195250.hg.1 | NR_125345.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900195271.hg.1 | ENST00000596746.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195271.hg.1 | ENST00000598892.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195271.hg.1 | NR_125345.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900195271.hg.1 | TC1900011295.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900195271.hg.1 | ENST00000596746.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195271.hg.1 | ENST00000598892.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195271.hg.1 | NR_125345.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900195275.hg.1 | ENST00000596746.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195275.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195275.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195275.hg.1 | NR_125345.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900195275.hg.1 | TC1900011295.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900195275.hg.1 | ENST00000596746.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195275.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195275.hg.1 | ENST00000601562.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900195275.hg.1 | NR_125345.1 | Refseq |
