Assay Details
| Gene Symbol: TP53TG3HP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_034019.2 | 1 - 2 | 839 | 100 | ||
| GenBank mRNA | AA401542.1 | - | 1 - 2 | 100 | ||
| AA824251.1 | - | 1 - 2 | 100 | |||
| BC038761.1 | - | 1 - 2 | 131 | 100 | ||
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30528840 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09989801 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16891495 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30528783 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30528776 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30528823 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16891468 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16891474 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09989800 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09989776 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16891505 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30528811 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16891538 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16891491 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16891529 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16891487 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30528819 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09989785 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09989779 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09989788 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30528755 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30528815 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09989803 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16891482 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30528760 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09989794 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09989772 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30528837 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30528766 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09989790 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09989808 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09989774 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09989798 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16891512 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30528770 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09989791 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09989771 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30528769 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30528836 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16891515 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30528812 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30528781 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30528747 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09989792 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09989781 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16891518 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16891514 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09989787 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09989784 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30528801 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09989809 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30528800 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09989793 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30528763 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30528817 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30528816 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30528825 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09989789 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 390518 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_32_P222460 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 18994 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 51804 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P222460 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_32_P222460 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P222460 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 237730_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010153.hg.1 | ENST00000562591.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010153.hg.1 | ENST00000562591.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010153.hg.1 | ENST00000562591.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010153.hg.1 | NR_034018.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010153.hg.1 | NR_034018.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010153.hg.1 | NR_034018.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010153.hg.1 | NR_034018.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010153.hg.1 | NR_034019.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010153.hg.1 | NR_034019.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010153.hg.1 | NR_034019.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010153.hg.1 | ENST00000562591.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010153.hg.1 | ENST00000562591.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010153.hg.1 | ENST00000562591.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010153.hg.1 | NR_034018.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010153.hg.1 | NR_034018.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010153.hg.1 | NR_034018.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010153.hg.1 | NR_034018.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010153.hg.1 | NR_034019.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010153.hg.1 | NR_034019.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010153.hg.1 | NR_034019.2 | Refseq |
| Human Genome U133 Plus 2.0 Array | 237730_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 237730_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 237730_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 237730_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 237730_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 237730_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 237730_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 237730_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 237730_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 237730_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 237730_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 237730_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 237730_at7 | - | - |
| Human Genome U95 Set | 63039_PM_at | - | - |
| Human Genome U95 Set | 63038_r_at | - | - |
| Human Genome U95 Set | 63039_at | - | - |
| Human Genome U95 Set | 63037_PM_s_at | - | - |
| Human Genome U95 Set | 63037_s_at | - | - |
| Human Genome U95 Set | 63038_PM_r_at | - | - |
| Human Genome U133 Set | 237730_PM_at | - | - |
| Human Genome U133 Set | 237730_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A053718 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A053718 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 715423 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3252273 | NR_034018.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3252273 | AI653002.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3175815 | NR_034018.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3175815 | NR_034019.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3175815 | AA401542.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3175815 | AA824251.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3175815 | AI653002.1 | human_transcriptome |

TP53TG3HP
Genomic Map