Assay Details
| Gene Symbol: HPSE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098540.2 | NP_001092010.1 | 12 | 2985 | 144 | |
| NM_001166498.2 | NP_001159970.1 | 11 | 2801 | 144 | ||
| NM_001199830.1 | NP_001186759.1 | 11 | 2743 | 144 | ||
| NM_006665.5 | NP_006656.2 | 13 | 3023 | 144 | ||
| GenBank mRNA | AK090910.1 | - | 1 | 794 | 144 | |
More Information
Set Membership: |
|

HPSE
Genomic Map