Assay Details
| Gene Symbol: HNRNPA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194247.2 | NP_919223.1 | 11 | 3216 | 150 | |
| XM_005246381.4 | XP_005246438.1 | 11 | 3122 | 150 | ||
| XM_006712365.2 | XP_006712428.1 | 10 | 3075 | 150 | ||
More Information
Updated Mapping Information
Set Membership: |
|

HNRNPA3
Genomic Map