Assay Details
| Gene Symbol: PTTG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282382.1 | NP_001269311.1 | 1 | 356 | 145 | |
| GenBank mRNA | AJ223953.1 | - | 1 | 58 | 145 | |
| BI827526.1 | - | 1 | 356 | 145 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AJ223953.1, NM_001282382.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
DNA repair
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
mitotic nuclear division
spermatogenesis
negative regulation of endopeptidase activity
anaphase-promoting complex-dependent catabolic process
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
homologous chromosome segregation
chromosome organization
cell division
transcription factor activity, sequence-specific DNA binding
cysteine-type endopeptidase inhibitor activity
protein binding
SH3 domain binding
peptidase inhibitor activity
DNA repair
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
mitotic nuclear division
spermatogenesis
negative regulation of endopeptidase activity
anaphase-promoting complex-dependent catabolic process
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
homologous chromosome segregation
chromosome organization
cell division
transcription factor activity, sequence-specific DNA binding
cysteine-type endopeptidase inhibitor activity
protein binding
SH3 domain binding
peptidase inhibitor activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000352433.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000352433.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000352433.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000352433.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000352433.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000352433.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000352433.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000352433.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000352433.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000393964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000393964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000393964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000393964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000393964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000393964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000393964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000393964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000393964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000393964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000393964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000393964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000517480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000517480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000517480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000517480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000517480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000519287.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000519287.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000519287.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000519287.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000519287.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000519287.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000519287.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000519287.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000520452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000520452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000520452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000520452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000520452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000520452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000520452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000520452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000523659.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000523659.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000523659.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000523659.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000523659.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000523659.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000524244.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000524244.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000524244.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | ENST00000524244.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_001282382.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_001282382.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_001282382.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_001282382.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_001282382.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_001282382.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_001282382.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_001282382.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_001282382.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_001282382.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_001282382.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_001282382.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_001282383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_001282383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_001282383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_001282383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_001282383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_001282383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_001282383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_001282383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_001282383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_004219.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_004219.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_004219.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_004219.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_004219.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_004219.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_004219.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_004219.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009281.hg.1 | NM_004219.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000352433.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000352433.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000352433.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000352433.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000352433.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000352433.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000352433.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000352433.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000352433.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000393964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000393964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000393964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000393964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000393964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000393964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000393964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000393964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000393964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000393964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000393964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000393964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000517480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000517480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000517480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000517480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000517480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000519287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000519287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000519287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000519287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000519287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000519287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000519287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000519287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000520452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000520452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000520452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000520452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000520452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000520452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000520452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000520452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000523659.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000523659.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000523659.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000523659.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000523659.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000523659.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000524244.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000524244.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000524244.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | ENST00000524244.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_001282382.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_001282382.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_001282382.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_001282382.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_001282382.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_001282382.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_001282382.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_001282382.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_001282382.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_001282382.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_001282382.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_001282382.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_001282383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_001282383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_001282383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_001282383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_001282383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_001282383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_001282383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_001282383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_001282383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_004219.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_004219.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_004219.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_004219.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_004219.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_004219.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_004219.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_004219.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009281.hg.1 | NM_004219.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500164839.hg.1 | ENST00000393964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500164839.hg.1 | ENST00000519287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500164839.hg.1 | NM_001282382.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500164839.hg.1 | ENST00000393964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500164839.hg.1 | ENST00000519287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500164839.hg.1 | NM_001282382.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500164839.hg.1 | TC0500009281.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500164840.hg.1 | ENST00000393964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500164840.hg.1 | NM_001282382.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500164840.hg.1 | ENST00000393964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500164840.hg.1 | NM_001282382.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500164840.hg.1 | TC0500009281.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500164837.hg.1 | ENST00000393964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500164837.hg.1 | NM_001282382.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500164837.hg.1 | ENST00000393964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500164837.hg.1 | NM_001282382.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500164837.hg.1 | TC0500009281.hg.1 | Clariom D TC |

PTTG1
Genomic Map