Assay Details
| Gene Symbol: IGLV3-19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | L43092.2 | - | 1 | 233 | 150 | |
| Gene Symbol: IGLJ3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB001733.1 | - | 618 | 150 | ||
| AB001736.1 | - | 618 | 150 | |||
| AB014341.1 | - | 645 | 150 | |||
| L35918.1 | - | 2 | 294 | 150 | ||
| Gene Symbol: IGLL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB064186.1 | - | 1 | 303 | 150 | |
| AB064212.1 | - | 1 | 303 | 150 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human 2 cDNA Microarray (G4101A) | 3054669 | - | - |
| Agilent Human 2 cDNA Microarray (G4101A) | 3054669 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 7981731 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7981730 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 216365_PM_x_at | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 211881_PM_x_at | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 211798_PM_x_at | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 216853_PM_x_at | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 214677_PM_x_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006797.hg.1 | ENST00000520756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006797.hg.1 | ENST00000520756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010481.hg.1 | ENST00000520119.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010481.hg.1 | ENST00000520119.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006816.hg.1 | ENST00000519345.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006789.hg.1 | ENST00000523713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006789.hg.1 | ENST00000523713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010113.hg.1 | ENST00000561990.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010113.hg.1 | ENST00000561990.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009213.hg.1 | ENST00000390309.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009213.hg.1 | ENST00000390309.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009213.hg.1 | ENST00000390309.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009213.hg.1 | ENST00000390309.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006816.hg.1 | ENST00000519345.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006797.hg.1 | ENST00000520756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006797.hg.1 | ENST00000520756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006789.hg.1 | ENST00000523713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006789.hg.1 | ENST00000523713.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010481.hg.1 | ENST00000520119.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010481.hg.1 | ENST00000520119.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010113.hg.1 | ENST00000561990.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010113.hg.1 | ENST00000561990.3 | Ensembl |
| Human Genome U133 Plus 2.0 Array | 203174_s_at | - | - |
| Human Genome U133 Plus 2.0 Array | 216365_PM_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 216846_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 216853_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 211798_PM_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 216852_PM_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 211798_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 211881_PM_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 214677_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 216853_PM_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 215984_s_at | - | - |
| Human Genome U133 Plus 2.0 Array | 216851_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 211881_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 215984_PM_s_at | - | - |
| Human Genome U133 Plus 2.0 Array | 216852_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 216853_PM_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 216853_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 214677_PM_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 216365_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 203174_PM_s_at | - | - |
| Human Genome U133 Plus 2.0 Array | 216851_at | - | - |
| Human Genome U133 Plus 2.0 Array | 216846_at | - | - |
| Human Genome U133A 2.0 Array | 216365_PM_x_at | - | - |
| Human Genome U133A 2.0 Array | 214677_PM_x_at | - | - |
| Human Genome U133A 2.0 Array | 211881_PM_x_at | - | - |
| Human Genome U133A 2.0 Array | 211798_x_at | - | - |
| Human Genome U133A 2.0 Array | 211798_PM_x_at | - | - |
| Human Genome U133A 2.0 Array | 216853_PM_x_at | - | - |
| Human Genome U133A 2.0 Array | 214677_x_at | - | - |
| Human Genome U133A 2.0 Array | 216846_at | - | - |
| Human Genome U133A 2.0 Array | 215984_s_at | - | - |
| Human Genome U133A 2.0 Array | 203174_PM_s_at | - | - |
| Human Genome U133A 2.0 Array | 215984_PM_s_at | - | - |
| Human Genome U133A 2.0 Array | 216365_x_at | - | - |
| Human Genome U133A 2.0 Array | 203174_s_at | - | - |
| Human Genome U133A 2.0 Array | 216853_x_at | - | - |
| Human Genome U133A 2.0 Array | 211881_x_at | - | - |
| Human Genome U133A 2.0 Array | 216846_PM_at | - | - |
| AffymetrixPathwork Diagnostics Pathchip | pw567208 | - | - |
| AffymetrixPathwork Diagnostics Pathchip | pw502325 | - | - |
| AffymetrixPathwork Diagnostics Pathchip | pw585174 | - | - |
| Human Genome U95 Set | 35765_at | - | - |
| Human Genome U95 Set | 35765_PM_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214677_x_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 211798_x_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 211881_x_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214677_x_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 211881_x_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 211881_x_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 211881_x_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214677_x_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 211881_x_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 216853_x_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 216853_x_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 211798_x_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 211798_x_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214677_x_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 216853_x_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 211881_x_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 211798_x_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 211798_x_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 216853_x_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 211881_x_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214677_x_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214677_x_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 211881_x_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 211798_x_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214677_x_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 211881_x_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214677_x_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 216853_x_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 216853_x_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214677_x_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 211881_x_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 216853_x_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 211798_x_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 211798_x_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 216853_x_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 216853_x_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 211798_x_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 216853_x_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 211798_x_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 211798_x_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214677_x_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 216853_x_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 211881_x_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214677_x_at3 | - | - |
| Human Genome U95 Set | 73327_PM_r_at | - | - |
| Human Genome U95 Set | 73327_r_at | - | - |
| Human Genome U133 Set | 216846_PM_at | - | - |
| Human Genome U133 Set | 216365_PM_x_at | - | - |
| Human Genome U133 Set | 211798_PM_x_at | - | - |
| Human Genome U133 Set | 215984_s_at | - | - |
| Human Genome U133 Set | 211881_PM_x_at | - | - |
| Human Genome U133 Set | 215984_PM_s_at | - | - |
| Human Genome U133 Set | 203174_s_at | - | - |
| Human Genome U133 Set | 203174_PM_s_at | - | - |
| Human Genome U133 Set | 216365_x_at | - | - |
| Human Genome U133 Set | 211798_x_at | - | - |
| Human Genome U133 Set | 214677_x_at | - | - |
| Human Genome U133 Set | 216853_PM_x_at | - | - |
| Human Genome U133 Set | 216853_x_at | - | - |
| Human Genome U133 Set | 216846_at | - | - |
| Human Genome U133 Set | 211881_x_at | - | - |
| Human Genome U133 Set | 214677_PM_x_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 125743 | - | - |
| ABI Human Genome Survey Microarray Version 2.0 | 181039 | - | - |

IGLV3-19
Genomic Map